Skip to Content
Merck
All Photos(3)

Key Documents

483044

Sigma-Aldrich

Potassium dichromate

99.98% trace metals basis

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

Assay

99.98% trace metals basis

form

crystalline

reaction suitability

reagent type: oxidant

technique(s)

cell culture | mammalian: suitable

impurities

≤150.0 ppm Trace Metal Analysis

mp

398 °C (lit.)

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium dichromate can be used as an oxidizing agent for the synthesis of polymers such as poly-O-phenylenediamine and polyanilines via oxidative polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of fluorine-substituted polyanilines
A Cihaner and A.M Onal
European Polymer Journal, 37, 1767-1772 (2001)
Chemical polymerization kinetics of poly-o-phenylenediamine and characterization of the obtained polymer in aqueous hydrochloric acid solution using K2Cr2O7 as oxidizing agent
SM Sayyah, et al.
International Journal of Polymer Science, 2014 (2014)
Peter A Siver et al.
American journal of botany, 102(6), 921-941 (2015-06-24)
Heterokont algae of the class Synurophyceae, characterized by distinctive siliceous scales that cover the surface of the cell, are ecologically important in inland waters, yet their evolutionary history remains enigmatic. We explore phylogenetic relationships within this group of algae relative
Bour-Jr Wang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 110(12), 775-779 (2012-01-18)
Occupational dermatitis among cement workers is a major occupational health concern. The two most important occupational hazards for cement workers are irritant and allergic cement contact dermatitis. The objective of this study was to investigate the severity of occupational cement
Shafaqat Ali et al.
Environmental toxicology and chemistry, 32(10), 2234-2239 (2013-06-19)
A hydroponic experiment was carried out to examine the effect of hydrogen sulfide (H2 S) in alleviating chromium (Cr) stress in barley. A 2-factorial design with 6 replications was selected, including 3 levels of NaHS (0 μM, 100 μM, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service