Skip to Content
Merck
All Photos(1)

Key Documents

529508

Sigma-Aldrich

Potassium chromate

≥99.95% trace metals basis

Synonym(s):

Chromate of potash, Dipotassium chromate, Dipotassium dioxido(dioxo)chromium, Dipotassium monochromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.7 (vs air)

Assay

≥99.95% trace metals basis

form

solid

impurities

≤500.0 ppm Trace Metal Analysis

mp

971 °C (lit.)

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lirong Liang et al.
Journal of neurochemistry, 154(4), 372-388 (2019-11-11)
Sevoflurane is one of the most widely used anesthetics with recent concerns rising about its pediatric application. The synaptic toxicity and mechanisms underlying its long-term cognition impairment remain unclear. In this study, we investigated the expression and roles of homeodomain
Xue Zhou et al.
Toxicology and applied pharmacology, 236(1), 78-84 (2009-04-18)
Occupational exposure to nickel (Ni), chromium (Cr), and arsenic (As) containing compounds has been associated with lung cancer and other adverse health effects. Their carcinogenic properties may be attributable in part, to activation and/or repression of gene expression induced by
Cecylia Wardak
Journal of hazardous materials, 186(2-3), 1131-1135 (2010-12-21)
A new polyvinylchloride membrane sensor for Pb(2+) with solid contact based on ionic liquid has been prepared. The electrode shows a Nernstian response for lead ions over a wide concentration range (1×10(-8) to 1×10(-1) mol L(-1)) and the slope of
Subhendra N Mattagajasingh et al.
Journal of applied toxicology : JAT, 28(8), 987-997 (2008-07-11)
Hexavalent chromium [Cr(VI)] compounds are Group-I human carcinogens. Cr(VI)-induced DNA-protein crosslinks (DPCs) have been implicated in the mutagenic and carcinogenic effects of Cr(VI). Although multiple mechanisms have been suggested for Cr(VI)-induced DNA-protein crosslinking, the mechanism of formation of DNA-protein crosslinks
Nejla Soudani et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 63(6), 541-548 (2010-05-25)
Chromium, a major environmental pollutant, is known for its wide toxic manifestations. The present experiment pertains to the protective role of selenium (Se) against K(2)Cr(2)O(7)-induced hepatotoxicity. Female Wistar rats were divided into four groups of six each: group I served

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service