Skip to Content
Merck
All Photos(3)

Documents

228621

Sigma-Aldrich

Lead(II) nitrate

ACS reagent, ≥99.0%

Synonym(s):

Lead dinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: lead

impurities

≤0.005% insolubles

pH

3-4 (20 °C, 50 g/L)

mp

470 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.005%
Na: ≤0.02%

SMILES string

[PbH2++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2

InChI key

RLJMLMKIBZAXJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Several new polymeric complexes of lead with dialkylamino derivatives of squaric acid were produced to study their coordination modes as possible prototypes for studies of lanthanide complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of Cell Complexity in the Filamentous Cyanobacterium Mastigocladus laminosus by Transmission Electron Microscopy (TEM).
Nurnberg DJ, et al.
Bio-protocol, 4(23) (2014)
Crown ether complexes of lead (II) nitrate. Crystal structures of the 12-crown-4, 15-crown-5, benzo-15-crown-5 and 18-crown-6 complexes.
Rogers RD and Bond AH.
Inorgorganica Chimica Acta, 192(2), 163-171 (1992)
Lincoln A. Hall et al.
Inorganic chemistry, 36(14), 3096-3101 (1997-07-02)
Reaction of three different 1-(dialkylamino) derivatives of 3,4-dihydroxycyclobut-3-ene-1,2-dione (squaric acid) with lead(II) nitrate in aqueous solution yielded polymeric complexes which have been characterized by single-crystal X-ray analysis. Pb[(CH(3))(2)NC(4)O(3)](2)(OH(2))(2).H(2)O (1) produced from 1-(dimethylamino)-2-methoxycyclobutenedione crystallizes in the triclinic space group P&onemacr;, with
Solid-state lead-207 NMR of lead (II) nitrate: Localized heating effects at high magic angle spinning speeds.
van Gorkom L, et al.
Magnetic Resonance in Chemistry, 33(10), 791-795 (1995)
Lewis-base adducts of lead (II) compounds. VII. Synthetic and structural studies of some 1: 1 adducts of linear polyamines with lead (II) nitrate.
Harrowfield JM, et al.
Australian Journal of Chemistry, 49(10), 1029-1042 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service