Skip to Content
Merck
All Photos(3)

Key Documents

S4547

Sigma-Aldrich

Sodium bromide

BioXtra, ≥99.0%

Synonym(s):

NaBr, natrium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050404
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99.0%
solubility:
H2O: 1 M, clear, colorless

vapor pressure

1 mmHg ( 806 °C)

Quality Level

product line

BioXtra

Assay

≥99.0%

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

mp

755 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ba: ≤0.002%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.1%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium bromide has been used:
  • to dialyze DNA solutions to keep the DNA helix intact
  • to maintain the relative humidity of edible film samples, stored in desiccator
  • as reference standard to confirm bromide ion retention time

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical and structural characterisation of zein and chitosan edible films using nanotechnology tools
Escamilla GM, et al.
International Journal of Biological Macromolecules, 61, 196-203 (2013)
Indirect Determination of Process Impurity Cetrimonium Bromide in Neisseria meningitidis A/C/Y/W-135-DT Conjugate Vaccine by HPAEC-PAD Method
Gudlavallet SK, et al.
International Journal of Biological Macromolecules, 1 (2016)
Confinement of DNA in water-in-oil microemulsions
Swami A, et al.
Langmuir, 24(20), 11828-11833 (2008)
David G Levitt et al.
Journal of applied physiology (Bethesda, Md. : 1985), 109(3), 786-795 (2010-06-19)
Measurement of body composition changes following bariatric surgery is complicated because of the difficulty of measuring body fat in highly obese individuals that have increased photon absorption and are too large for the standard dual-energy X-ray absorptiometry (DXA) table. We
Julian Holley et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 84(5 Pt 2), 056110-056110 (2011-12-21)
Excitation waves on a subexcitable Belousov-Zhabotinsky (BZ) substrate can be manipulated by chemical variations in the substrate and by interactions with other waves. Symbolic assignment and interpretation of wave dynamics can be used to perform logical and arithmetic computations. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service