Skip to Content
Merck
All Photos(1)

Key Documents

P9564

Sigma-Aldrich

Phleomycin

from Streptomyces verticillus, powder, DNA replication inhibitor

Synonym(s):

FT-0697305

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Phleomycin from Streptomyces verticillus, powder

form

powder

Quality Level

shelf life

2 yr

color

blue

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

InChI

1S/C51H75N17O21S2/c1-16-29(65-41(68-38(16)54)21(8-27(53)72)60-9-20(52)39(55)78)44(82)67-31(34(22-10-58-15-61-22)88-49-50(5,36(76)33(74)25(11-69)87-49)89-47-37(77)51(85,48(57)84)35(75)26(12-70)86-47)45(83)62-18(3)32(73)17(2)42(80)66-30(19(4)71)43(81)59-7-6-28-63-24(14-90-28)46-64-23(13-91-46)40(56)79/h10,13,15,17-18,20-21,24-26,30-37,47,49,60,69-70,73-77,85H,6-9,11-12,14,52H2,1-5H3,(H2,53,72)(H2,55,78)(H2,56,79)(H2,57,84)(H,58,61)(H,59,81)(H,62,83)(H,66,80)(H,67,82)(H2,54,65,68)/t17?,18?,20?,21?,24?,25-,26?,30?,31?,32?,33+,34?,35-,36-,37-,47-,49-,50-,51+/m0/s1

InChI key

QRBLKGHRWFGINE-UGWAGOLRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phleomycin is a glycopeptide antibiotic and is a member of the bleomycin family.

Application

Phleomycin from Streptomyces verticillus has been used as an antibiotic in the screening of transformants of Trypanosoma brucei. It has also been used as an antibiotic in the maintenance of Escherichia coli.

Biochem/physiol Actions

Phleomycin can bind and intercalate with the DNA that leads to DNA strand breaks. It acts as an inhibitor of DNA replication.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

James B Wilson et al.
Mutation research, 689(1-2), 12-20 (2010-05-11)
The Fanconi anaemia (FA) FANCG protein is an integral component of the FA nuclear core complex that is required for monoubiquitylation of FANCD2. FANCG is also part of another protein complex termed D1-D2-G-X3 that contains FANCD2 and the homologous recombination
Appolinaire Djikeng et al.
Methods in molecular biology (Clifton, N.J.), 265, 73-83 (2004-04-23)
Trypanosoma brucei, a flagellate protozoa of the family Trypanosomatidae, has become one of the model systems for unicellular pathogens to study fundamentally important biological phenomena. Currently, the method of choice to examine gene function in these organisms is RNA interference
C H He et al.
Canadian journal of microbiology, 42(12), 1263-1266 (1996-12-01)
The antibiotic bleomycin is used as an anticancer agent for treating a variety of tumours. The antitumour effect of bleomycin is related to its ability to produce lesions such as apurinic/apyrimidinic sites and single- and double-strand breaks in the cellular
M J Sleigh et al.
Mutation research, 42(2), 181-190 (1977-02-01)
Sulphydryl-mediated DNA breakage, which is induced by the antibiotic phleomycin in vitro, has been found to contribute significantly to the DNA damage produced by phleomycin in Escherichia coli. The effect of pleomycin was inhibited in vivo, as in vitro, by
L F Povirk et al.
Biochemistry, 20(3), 665-671 (1981-02-03)
The kinetics and mechanism of binding of Cu-(II).bleomycin, Fe(III).bleomycin, and Cu(II).phleomycin to DNA were studied by using fluorometry, equilibrium dialysis, electric dichroism, and temperature-jump and stopped-flow spectrophotometry. The affinity of Cu(II).bleomycin for DNA was greater than that of metal-free bleomycin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service