Skip to Content
Merck
All Photos(5)

Key Documents

S4641

Sigma-Aldrich

Sodium citrate tribasic dihydrate

ACS reagent, ≥99.0%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
grade:
ACS reagent
solubility:
water: soluble 29.4 g/L at 20 °C (completely)

grade

ACS reagent

Quality Level

Agency

suitable for SM 4500 - NH3

Assay

≥99.0%

impurities

≤0.003% Ammonia (NH3)
≤0.005% Insoluble matter

pH

7.0-9.0 (25 °C, 5% in solution)

mp

>300 °C (lit.)

solubility

water: soluble 29.4 g/L at 20 °C (completely)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Sodium citrate tribasic dihydrate can be used as a catalyst to synthesize:     
  • Fused O- and N-heterocyclic derivatives via multicomponent Knoevenagel-cyclocondensation of isatin/aldehyde/ acenaphthylene-1,2-dione, malononitrile, and various C−H activated acids.     
  • Chromeno[3′,4′:5,6]pyrano[2,3-d]pyrimidines by the condensation reaction of 4-hydroxycoumarin, aromatic aldehydes, and barbituric acid/thiobarbituric acid.

It can also be used as:      
A reducing agent to synthesize metal nanoparticles via sol-gel method using corresponding metal salt precursors.    
A controlling agent in the synthesis of single-crystal of coordination polymer [Co(H2O)2Ni(CN)4 4H2O].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyungbin Kim et al.
Polymers, 11(4) (2019-04-19)
Complex coacervation is an emerging liquid/liquid phase separation (LLPS) phenomenon that behaves as a membrane-less organelle in living cells. Yet while one of the critical factors for complex coacervation is temperature, little analysis and research has been devoted to the
Wei Yu et al.
Nature communications, 11(1), 5239-5239 (2020-10-18)
The alternative non-homologous end-joining (NHEJ) pathway promotes DNA double-strand break (DSB) repair in cells deficient for NHEJ or homologous recombination, suggesting that it operates at all stages of the cell cycle. Here, we use an approach in which DNA breaks
Ben Ouyang et al.
Nature materials, 19(12), 1362-1371 (2020-08-12)
Nanoparticle delivery to solid tumours over the past ten years has stagnated at a median of 0.7% of the injected dose. Varying nanoparticle designs and strategies have yielded only minor improvements. Here we discovered a dose threshold for improving nanoparticle tumour
Xian Zhang et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(6), 1690-1700 (2019-04-13)
Mast cell (MC) deficiency in KitW-sh/W-sh mice and inhibition with disodium chromoglycate (DSCG) or ketotifen reduced obesity and diabetes in mice on a high-cholesterol (1.25%) Western diet. Yet, Kit-independent MC-deficient mice and mice treated with DSCG disproved MC function in
Andrés Di Paolo et al.
RNA (New York, N.Y.), 26(11), 1637-1653 (2020-08-05)
Programmed cell death 4 (PDCD4) protein is a tumor suppressor that inhibits translation through the mTOR-dependent initiation factor EIF4A, but its functional role and mRNA targets in neurons remain largely unknown. Our work identified that PDCD4 is highly expressed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service