Skip to Content
Merck
All Photos(3)

Key Documents

310506

Sigma-Aldrich

Sodium bromide

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050404
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder, crystals or granules
solubility:
ethanol: soluble(lit.)
water: soluble(lit.)

grade

ACS reagent

Agency

suitable for EPA 300

vapor pressure

1 mmHg ( 806 °C)

Assay

≥99.0%

form

powder, crystals or granules

impurities

≤0.005% insolubles

pH

5.0-8.8 (25 °C, 5%)

mp

755 °C (lit.)

solubility

ethanol: soluble(lit.)
water: soluble(lit.)

anion traces

bromate (BrO3-): ≤0.001%
chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.002%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
K: ≤0.1%
Mg: ≤0.001%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium Bromide is an inorganic compound that is commonly used as a source of bromide ions as a nucleophile or nucleophilic catalyst. It is also used as a precursor to reactive electrophilic bromine species, which are useful for a variety of organic transformations.

Application

Sodium Bromide can be used as a catalyst in TEMPO-mediated oxidation reactions. It is also used to grow the single crystal of γ-glycine by the slow cooling method.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium bromide catalysed one-pot synthesis of tetrahydrobenzo [b] pyrans via a three-component cyclocondensation under microwave irradiation and solvent free conditions.
Devi I and Bhuyan PJ.
Tetrahedron Letters, 45(47), 8625-8627 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 996-996 (1994)
Sergiu Coseri et al.
Carbohydrate polymers, 116, 9-17 (2014-12-03)
Unoxidized and carboxylated pullulan (obtained by pullulan oxidation using TEMPO-sodium hypochlorite-sodium bromide system) have been used as mediators for the silver nanoparticles formation (AgNPs), under environment-friendly conditions: using aqueous solutions, room temperature and notably, by using both mediators as reducing
Ling Yue et al.
Applied biochemistry and biotechnology, 175(7), 3557-3570 (2015-02-05)
A novel aqueous two-phase system (ATPS) only containing Gemini surfactant ethanediyl-1,2-bis(dodecyldimethylammonium bromide) and an inorganic salt sodium bromide was designed, and the physicochemical properties of the ATPS were investigated systematically. The results have shown that the coexisting two phases, one
Density, viscosity, and electrolytic conductivity of concentrated aqueous electrolyte solutions at several temperatures. Alkaline-earth chlorides, lanthanum chloride, sodium chloride, sodium nitrate, sodium bromide, potassium nitrate, potassium bromide, and cadmium nitrate.
Isono T.
Journal of Chemical and Engineering Data, 29(1), 45-52 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service