Skip to Content
Merck
All Photos(1)

Key Documents

PHR1691

Supelco

4-Epianhydrotetracycline hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Epianhydrotetracycline hydrochloride, (4R,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N2O7 · HCl
CAS Number:
Molecular Weight:
462.88
Beilstein:
3840711
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. E0400000
traceable to USP 1236506

API family

chlortetracycline, tetracycline

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].[H][C@@]12Cc3c(C)c4cccc(O)c4c(O)c3C(=O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@@H]2N(C)C

InChI

1S/C22H22N2O7.ClH/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H/t11-,16+,22-;/m0./s1

InChI key

XBSQEFHDCDFNJU-MOMXNFOMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Epianhydrotetracycline hydrochloride is a degradation product of tetracyclines which are used as veterinary medicines.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

4-Epianhydrotetracycline hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in commercial bulk and pharmaceutical products by high performance liquid chromatography (HPLC).
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6863 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A high performance liquid chromatographic system for the analysis of tetracycline drug standards, analogs, degradation products and other impurities
Mack GD and Ashworth RB
Journal of Chromatographic Science, 16(3), 93-101 (1978)
4-Epianhydrotetracycline
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 6636-6636 (2013)
Emad M Hussien
Biomedical chromatography : BMC, 28(9), 1278-1283 (2014-03-13)
A novel reversed-phase HPLC method was developed and validated for the assay of tetracycline hydrochloride and the limit of 4-epianhydrotetracycline hydrochloride impurity in tetracycline hydrochloride commercial bulk and pharmaceutical products. The method employed L1 (3 µm, 150 × 4.6 mm) columns, a mobile phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service