Skip to Content
Merck
All Photos(1)

Documents

37921

Supelco

4-Epianhydrotetracycline hydrochloride

VETRANAL®, analytical standard

Synonym(s):

(4R,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N2O7 · HCl
CAS Number:
Molecular Weight:
462.88
Beilstein:
3840711
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Cl[H].[H][C@@]12Cc3c(C)c4cccc(O)c4c(O)c3C(=O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@@H]2N(C)C

InChI

1S/C22H22N2O7.ClH/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H/t11-,16+,22-;/m0./s1

InChI key

XBSQEFHDCDFNJU-MOMXNFOMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martha Ines Burgos et al.
Biological & pharmaceutical bulletin, 34(8), 1301-1306 (2011-08-02)
Tetracycline (TC) derivatives are extensively used as antibiotics in human and animal medicine and, very recently, they have been screened as anti-amyloidogenic drugs. Anhydrotetracycline (AHTC) is one of the major degradation products of TC that has been linked to several
Markus W Obrist et al.
Plasmid, 68(1), 33-42 (2012-03-27)
In Yersinia, the most commonly used expression vectors for genetic studies such as gene complementation do not effectively allow for both induction and repression of gene expression. Additionally, there is no expression system available that can be induced in bacteria
Ute Seidel et al.
The journal of physical chemistry. B, 111(21), 6006-6014 (2007-05-08)
Molecular-dynamics simulations have been used to investigate the mechanism of induction of a mutant (revTetR) of the tetracycline repressor protein (TetR) that shows the reverse phenotype (i.e., it is induced in the absence of tetracyclines and not in their presence).
G B Zavilgelsky et al.
Biochemistry. Biokhimiia, 73(8), 906-911 (2008-09-09)
Anti-restriction proteins ArdA and Ocr are specific inhibitors of type I restriction-modification enzymes. The IncI1 transmissible plasmid ColIb-P9 ardA and bacteriophage T7 0.3(ocr) genes were cloned in pUC18 vector. Both ArdA (ColIb-P9) and Ocr (T7) proteins inhibit both restriction and
Samuel L Hutson et al.
PloS one, 5(11), e14057-e14057 (2010-12-03)
Molecular pathways regulating rapid proliferation and persistence are fundamental for pathogens but are not elucidated fully in Toxoplasma gondii. Promoters of T. gondii ribosomal proteins (RPs) were analyzed by EMSAs and ChIP. One RP promoter domain, known to bind an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service