Skip to Content
Merck
All Photos(1)

Documents

PHR1195

Supelco

Tinidazole Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Methyl-4(5)-nitroimidazole, 2-METHYL-5-NITROIMIDAZOLE, Metronidazole Impurity A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
Beilstein:
4032
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000087
traceable to USP 1667530

API family

tinidazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

251-255 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ncc([nH]1)[N+]([O-])=O

InChI

1S/C4H5N3O2/c1-3-5-2-4(6-3)7(8)9/h2H,1H3,(H,5,6)

InChI key

FFYTTYVSDVWNMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9011 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel D Rossouw et al.
Nuclear medicine and biology, 32(4), 385-394 (2005-05-10)
The inflammation- and infection-seeking properties of (131)I-labeled ornidazole, a 5-nitroimidazole derivative, have recently been reported. Whole-body images in rabbits showed a more rapid uptake in inflamed areas compared to (67)Ga. In the present study, two novel (123)I-labeled 2-methyl-4-nitroimidazole derivatives were
Fa-qing Ye et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 38(4), 260-263 (2003-08-02)
To study the synthesis and antibacterial activity of pyridonecarboxylic acid derivatives containing 2-methyl-5-nitroimidazol. Pyridonecarboxylic acid derivatives containing 2-methyl-5-nitroimidazol were synthesized primarily from 2-methyl-5-nitroimidazol, norfloxacin, ciprofloxacin, enoxacin via nucleophilic substitution and esterification. The antibacterial activity of the nine target compounds were
G Carignan et al.
Food additives and contaminants, 8(4), 467-475 (1991-07-01)
An unidentified metabolite of dimetridazole (DMZ), found in pig plasma, muscle and kidney, was shown by chromatography and spectroscopy to be 2-methyl-5-nitroimidazole (2-MNI), resulting from N-demethylation of DMZ. This route of degradation competes with the oxidation pathway previously described. The
Fa-qing Ye et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 40(2), 132-135 (2005-05-07)
To study the synthesis and antibacterial activity of ciprofloxacin derivatives. Ciprofloxacin derivatives were synthesized primarily from 2-methyl-5-nitroimidazol and ciprofloxacin through nucleophilic substitution. The antibacterial activity of the synthesized compounds were tested. Nine new compounds were synthesized. The structure of the
S Yang
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao, 30(3), 345-346 (2002-09-06)
The 2-methyl-5(4)-nitromidazole(BH) is an ampholyte with acid and base. It is an intermediate of metronidazole. The amphoteric dissociation constant of BH determined by equimolar UV-spectrophotometry. The acidic dissociation constant pKa is 9.64 +/- 0.04 and the basic dissociation constant pKb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service