Skip to Content
Merck
All Photos(1)

Documents

37919

Supelco

Anhydrotetracycline hydrochloride

VETRANAL®, analytical standard

Synonym(s):

(4S,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N2O7 · HCl
CAS Number:
Molecular Weight:
462.88
Beilstein:
4228856
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Cl[H].[H][C@@]12Cc3c(C)c4cccc(O)c4c(O)c3C(=O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@H]2N(C)C

InChI

1S/C22H22N2O7.ClH/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H/t11-,16-,22-;/m0./s1

InChI key

XBSQEFHDCDFNJU-WPJNXPDPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anhydrotetracycline (ATC) is a degradation product of tetracycline hydrochloride, a widely used antibiotic in animal breeding.

Application

The analytical standard can be used as follows:
  • Terahertz (THz) spectroscopy-based temperature-dependent quantitative and qualitative analysis of the antibiotic, tetracycline hydrochloride, and its three major metabolites
  • Simultaneous analysis of tetracycline hydrochloride and its degradation products in a pharmaceutical formulation by Resonance Raman Spectroscopy to study stress-induced stability of the drug
  • Development and validation of a liquid chromatography-tandem mass spectroscopy (LC-MS/MS) method to determine 21 antibiotic residues used as veterinary medicinal products in honey samples
  • Estimation of eight tetracycline antibiotics in various environmental water samples by dispersive liquid-liquid microextraction (DLLME) followed by an ultra-high pressure liquid chromatography (UHPLC)-tunable ultraviolet detection (TUV)

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susceptibility of Escherichia coli and Enterococcus faecium isolated from pigs and broiler chickens to tetracycline degradation products and distribution of tetracycline resistance determinants in E. coli from food animals
Sengelov G, et al.
Veterinary Microbiology, 95, 91-101 (2003)
Wurihan Wurihan et al.
Pathogens and disease, 77(9) (2020-02-06)
Chlamydiae are common, important pathogens for humans and animals alike. Despite recent advancement in genetics, scientists are still searching for efficient tools to knock out or knock down the expression of chromosomal genes. We attempted to adopt a dCas9-based CRISPR
James M Murithi et al.
Science translational medicine, 13(603) (2021-07-23)
The emergence and spread of Plasmodium falciparum resistance to first-line antimalarials creates an imperative to identify and develop potent preclinical candidates with distinct modes of action. Here, we report the identification of MMV688533, an acylguanidine that was developed following a
Jonathan W Kotula et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), 4838-4843 (2014-03-19)
The mammalian gut is a dynamic community of symbiotic microbes that interact with the host to impact health, disease, and metabolism. We constructed engineered bacteria that survive in the mammalian gut and sense, remember, and report on their experiences. Based
Wyndham W Lathem et al.
Science (New York, N.Y.), 315(5811), 509-513 (2007-01-27)
Primary pneumonic plague is transmitted easily, progresses rapidly, and causes high mortality, but the mechanisms by which Yersinia pestis overwhelms the lungs are largely unknown. We show that the plasminogen activator Pla is essential for Y. pestis to cause primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service