Skip to Content
Merck
All Photos(1)

Documents

T0600000

Tetracycline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Beilstein:
3844873
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tetracycline

manufacturer/tradename

EDQM

mp

220-223 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1

InChI key

XMEVHPAGJVLHIG-FMZCEJRJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tetracycline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuhiro Yomoda et al.
Journal of biomaterials applications, 29(10), 1385-1393 (2015-01-01)
Previous studies have reported the excellent biocompatibility of calcium phosphate cement. However, calcium phosphate cement needs further improvement in order for it to promote bone replacement and eventual bone substitution, as it exhibits slow biodegradability and thus remains in the
Joseph E Perosky et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(11), 1416-1423 (2014-08-05)
Heterotopic ossification (HO) is the abnormal formation of bone in soft tissue. Current diagnostics have low sensitivity or specificity to incremental progression of mineralization, especially at early time points. Without accurate and reliable early diagnosis and intervention, HO progression often
Que Chi Truong-Bolduc et al.
The Journal of infectious diseases, 209(9), 1485-1493 (2013-11-28)
Staphylococcus aureus colonizes skin in the presence of antimicrobial fatty acids and polyamines. The chromosomally encoded Tet38 efflux transporter confers resistance to tetracycline and fitness in abscesses, but its natural substrates and those of the Nor quinolone efflux pumps are
Yuqin Qiao et al.
Biomaterials, 35(25), 6882-6897 (2014-05-28)
Rapid development of zinc biology has broadened the applications of Zn-incorporated biomaterials to tissue engineering but also raised concerns about the long-term safety of released Zn(2+) ions. Clinical success hinges on the amount of incorporated zinc and subsequent optimized release
E Amram et al.
Antimicrobial agents and chemotherapy, 59(2), 796-802 (2014-11-19)
Mycoplasma bovis isolates with decreased susceptibilities to tetracyclines are increasingly reported worldwide. The acquired molecular mechanisms associated with this phenomenon were investigated in 70 clinical isolates of M. bovis. Sequence analysis of the two 16S rRNA-encoding genes (rrs3 and rrs4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service