Skip to Content
Merck
All Photos(1)

Key Documents

92360

Sigma-Aldrich

Chlorotrimethylsilane

puriss., ≥99.0% (GC)

Synonym(s):

TMCS, Trimethylchlorosilane, Trimethylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCl
CAS Number:
Molecular Weight:
108.64
Beilstein:
1209232
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

100 mmHg ( 25 °C)

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

752 °F

expl. lim.

6.4 %

refractive index

n20/D 1.387 (lit.)

bp

57 °C (lit.)

mp

−40 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

SMILES string

[H][C@@]12CC(C)(C)CC[C@]1(C)CC[C@]3(C)C2=CC[C@]4([H])[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C
C[Si](C)(C)Cl

InChI

1S/C3H9ClSi/c1-5(2,3)4/h1-3H3

InChI key

IJOOHPMOJXWVHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorotrimethylsilane is a chloro-organosilane compound mainly used for silylation reactions.

Application

  • Chlorotrimethylsilane can be a non-toxic alternative to mercuric chloride as activator in samarium-promoted cyclopropanation of both allylic and α-allenic alcohols.
  • It can be used in combination with hexamethyldisilazane for the protection of alcohols by forming trimethylsilyl ethers via silylation.
  • It is employed in chloromethylation of polysulfones (PSUs).
  • It activates lithium hydride to convert it into a hydride source for the reductive silylation of carbonyl compounds.
  • Chlorotrimethylsilane/lithium bromide forms an effective reagent for the conversion of alcohols to bromides.
  • Chlorotrimethylsilane/sodium iodide in acetonitrile is a better alternative to iodotrimethylsilane for the cleavage of esters, lactones, carbamates, and ethers. It can also be used for the conversion of alcohols to iodides.
  • Chlorotrimethylsilane along with silver or ammonium nitrate forms an efficient regioselective nitrating reagent for the ipso-nitration of arylboronic acids to form the corresponding nitroarenes.
  • Chlorotrimethylsilane along with sodium nitrite or nitrate can be used for deoximation reactions.

Other Notes

Silylating agent

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.H. Hergott et al.
Synthesis, 626-626 (1980)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
Chlorotrimethylsilane-sodium nitrite or nitrate. Efficient deoximating reagents for aldoximes and ketoximes.
Jong, Gun Lee and Ki, Hoon Kwak and Je, Pil Hwang
Tetrahedron Letters, 31(46), 6677-6680 (1990)
ipso-Nitration of Arylboronic Acids with Chlorotrimethylsilane- Nitrate Salts
Prakash GKS, et al.
Organic Letters, 6(13), 2205-2207 (2004)
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 4, 261-261 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service