Skip to Content
Merck
All Photos(2)

Key Documents

89595

Supelco

Chlorotrimethylsilane

for GC derivatization, LiChropur, ≥99.0% (GC)

Synonym(s):

TMCS, Trimethylchlorosilane, Trimethylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCl
CAS Number:
Molecular Weight:
108.64
Beilstein:
1209232
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

vapor density

3.7 (vs air)

vapor pressure

100 mmHg ( 25 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

752 °F

quality

LiChropur

expl. lim.

6.4 %

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.387 (lit.)

bp

57 °C (lit.)

mp

−40 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)Cl

InChI

1S/C3H9ClSi/c1-5(2,3)4/h1-3H3

InChI key

IJOOHPMOJXWVHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorotrimethylsilane is a silylation reagent used in the derivatization of triglycerides into volatile fatty esters by gas chromatography (GC).

Application

Learn more in the Product Information

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chlorotrimethylsilane as a reagent for gas chromatographic analysis of fats and oils.
Eras J, et al.
Journal of Chromatography A, 918(1), 227-232 (2001)
Ainara Sistiaga et al.
PloS one, 9(6), e101045-e101045 (2014-06-26)
Neanderthal dietary reconstructions have, to date, been based on indirect evidence and may underestimate the significance of plants as a food source. While zooarchaeological and stable isotope data have conveyed an image of Neanderthals as largely carnivorous, studies on dental
Nil Tandogan et al.
PloS one, 9(6), e101429-e101429 (2014-07-01)
We present an automated method for isolating pure bacterial cultures from samples containing multiple species that exploits the cell's own physiology to perform the separation. Cells compete to reach a chamber containing nutrients via a constriction whose cross-sectional area only
Kathleen A Vermeersch et al.
PloS one, 10(2), e0118262-e0118262 (2015-02-18)
Recently, multicellular spheroids were isolated from a well-established epithelial ovarian cancer cell line, OVCAR-3, and were propagated in vitro. These spheroid-derived cells displayed numerous hallmarks of cancer stem cells, which are chemo- and radioresistant cells thought to be a significant
Zachariah M Norman et al.
ACS nano, 8(7), 7513-7521 (2014-06-25)
We report the evolution of electrical transport and grain size during the sintering of thin films spin-cast from soluble phosphine and amine-bound, chloride-terminated cadmium selenide nanocrystals. Sintering of the nanocrystals occurs in three distinct stages as the annealing temperature is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service