Skip to Content
Merck
All Photos(1)

Documents

69478

Supelco

N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane

with 1% trimethylchlorosilane, for GC derivatization, LiChropur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12F3NOSi
CAS Number:
Molecular Weight:
199.25
Beilstein:
1941550
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

product name

N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, for GC derivatization, LiChropur

Quality Level

grade

for GC derivatization

Assay

≥97.0%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.378

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C

InChI

1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3

InChI key

MSPCIZMDDUQPGJ-UHFFFAOYSA-N

General description

Silylation can be performed by adding N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MTFA) during analysis of fluorometabolites via gas chromatography-mass spectrometry (GC-MS).

Application

Addition of Trimethylchlorosilane aids in the derivatization of amides, secondary amines and hindered hydroxy groups. Most volatile of the TMS derivatives often elutes at the solvent front of the GC.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Filamentous fungal biofilm for production of human drug metabolites.
Amadio, Jessica, Eoin Casey, and Cormac D. Murphy.
Applied Microbiology and Biotechnology, 97 (13), 5955-5963 (2013)
Stina K Lien et al.
Journal of chromatography. A, 1247, 118-124 (2012-06-15)
GC-MS analysis of silylated metabolites is a sensitive method that covers important metabolite groups such as sugars, amino acids and non-amino organic acids, and it has become one of the most important analytical methods for exploring the metabolome. Absolute quantitative
Harin Kanani et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 871(2), 191-201 (2008-06-10)
Metabolomics being the most recently introduced "omic" analytical platform is currently at its development phase. For the metabolomics to be broadly deployed to biological and clinical research and practice, issues regarding data validation and reproducibility need to be resolved. Gas
John A Bowden et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(27), 3237-3242 (2009-08-21)
Steroid derivatization was investigated by varying the experimental parameters (reagent, reaction time, and reaction temperature) to determine the optimal conditions for individual steroids, and for larger subsets. Three methods of derivatization enhancement were also investigated: the use of sonication, the
Hee-Kyung Jeon et al.
Journal of chromatography. A, 1131(1-2), 192-202 (2006-08-08)
A novel method has been developed to simultaneously determine and quantify seven organic UV filters employing liquid (solid)-liquid extraction, derivatization with N-methyl-N-(trimethylsilyl) trifluoroacetamide (MSTFA) and gas chromatography with mass spectrometric detection in various environmental matrices. The UV filters determined were:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service