Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
UNSPSC Code:
12352111
NACRES:
NA.22
PubChem Substance ID:
EC Number:
247-103-9
Beilstein/REAXYS Number:
2050024
MDL number:
Assay:
≥99%
Form:
liquid
grade
synthesis grade
Quality Level
assay
≥99%
form
liquid
refractive index
n20/D 1.384 (lit.)
bp
45-50 °C/14 mmHg (lit.)
density
0.969 g/mL at 25 °C (lit.)
functional group
fluoro
SMILES string
C[Si](C)(C)O\C(=N\[Si](C)(C)C)C(F)(F)F
InChI
1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3/b12-7+
InChI key
XCOBLONWWXQEBS-KPKJPENVSA-N
Related Categories
General description
N,O-Bis(trimethylsilyl)trifluoroacetamide is an organosilicon reagent for trimethylsilylation, deprotection, dehydrogenation, allylic alkylation, nitronate formation, and Claisen rearrangement.
BSTFA is a silylating agent used in the synthesis of 4-Quinolones.
BSTFA is a silylating agent used in the synthesis of 4-Quinolones.
Application
N,O-Bis(trimethylsilyl)trifluoroacetamide can be used as a derivatization reagent for the determination of the estrogens estrone and 17α-ethinylestradiol by gas chromatography-mass spectrometry.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
93.2 °F
flash_point_c
34 °C
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of 4-Quinolones: N, O-Bis (trimethylsilyl) acetamide-Mediated Cyclization with Cleavage of Aromatic C--O Bond
Pisa O, et al.
European Journal of Organic Chemistry, 2016, 2336-2350 (2016)
E Teixidó et al.
Journal of chromatography. A, 1135(1), 85-90 (2006-10-03)
A new, simple and selective method for the analysis of 5-hydroxymethylfurfural (HMF) in foods by gas chromatography coupled to mass spectrometry (GC-MS) is proposed. Several derivatising procedures based on the formation of an HMF silylated derivative using different reagents were
Hidenori Matsukami et al.
The Science of the total environment, 493, 672-681 (2014-07-06)
Chlorinated and non-halogenated organophosphorus flame retardants (OPFRs) including tris(2-chloroisopropyl) phosphate (TCIPP), diethylene glycol bis(di(2-chloroisopropyl) phosphate) (DEG-BDCIPP), triphenyl phosphate (TPHP), and bisphenol A bis(diphenyl phosphate) (BPA-BDPP) have been used increasingly as alternatives to polybrominated diphenyl ethers and other brominated flame retardants.
Ahmed I Rushdi et al.
SpringerPlus, 3, 253-253 (2014-06-14)
Propolis is a sticky material mixed by honeybees to utilize it in protecting their hives from infection by bacteria and fungi. The therapeutic properties of propolis are due to its chemical composition with bio-active compounds; therefore, researchers are interested in
Nyi Nyi Naing et al.
Journal of chromatography. A, 1423, 1-8 (2015-11-05)
Electro membrane extraction-solid-liquid phase microextraction (EME-SLPME) was developed for the first time to determine phenolic contaminants in water. The extraction system consisted of a solid/liquid interface that permitted a three-phase microextraction approach involving an aqueous sample (donor phase): an organic
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 155195-500G | 04061833498675 |
| 155195-25G | 04061838741950 |
| 155195-5G | 04061837789458 |
| 155195-100G | 04061837789441 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
