Skip to Content
Merck
All Photos(1)

Key Documents

683248

Sigma-Aldrich

(11bR)-2,6-Bis(diphenylphosphino)-N,N-dimethyldinaphtho[2,1-d:1′,2′-f]-1,3,2-dioxaphosphepin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H36NO2P3
CAS Number:
Molecular Weight:
727.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

245-249 °C

functional group

amine
phosphine

storage temp.

2-8°C

SMILES string

CN(C)P1Oc2c(cc3ccccc3c2-c4c(O1)c(cc5ccccc45)P(c6ccccc6)c7ccccc7)P(c8ccccc8)c9ccccc9

InChI

1S/C46H36NO2P3/c1-47(2)52-48-45-41(50(35-21-7-3-8-22-35)36-23-9-4-10-24-36)31-33-19-15-17-29-39(33)43(45)44-40-30-18-16-20-34(40)32-42(46(44)49-52)51(37-25-11-5-12-26-37)38-27-13-6-14-28-38/h3-32H,1-2H3

InChI key

JVTXDRKUFXCFCF-UHFFFAOYSA-N

Application

(11bR)-2,6-Bis(diphenylphosphino)-N,N-dimethyldinaphtho[2,1-d:1′,2′-f]-1,3,2-dioxaphosphepin-4-amine is a phosphine-phosphoramidite ligand, which can be used in:
  • The stereoselective preparation of aminoesters via rhodium-catalyzed enantioselective hydrogenation of dehydroaminoesters.
  • The Rh-catalyzed asymmetric hydrogenation of N-(α-arylvinyl) acetamides.

Legal Information

Sold under license from DSM for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of Triphosphorous Bidentate Phosphine-Phosphoramidite Ligands: Application in the Highly Enantioselective Hydrogenation of ortho-Substituted Aryl Enamides
Zhang W and Zhang X
Angewandte Chemie (International ed. in English), 118(33), 5641-5644 (2006)
Highly enantioselective hydrogenation of α-dehydroamino esters and itaconates with triphosphorous bidentate ligands and the unprecedented solvent effect thereof
Zhang W and Zhang X
The Journal of Organic Chemistry, 72(3), 1020-1023 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service