Skip to Content
Merck
All Photos(1)

Documents

89483

Sigma-Aldrich

1-Ethyl-3-methylimidazolium bromide

≥97.0% (T)

Synonym(s):

1-Ethyl-3-methyl-1H-imidazolium bromide, 1-Methyl-3-ethylimidazolium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BrN2
CAS Number:
Molecular Weight:
191.07
Beilstein:
5162084
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

form

crystals

SMILES string

[Br-].CCn1cc[n+](C)c1

InChI

1S/C6H11N2.BrH/c1-3-8-5-4-7(2)6-8;/h4-6H,3H2,1-2H3;1H/q+1;/p-1

InChI key

GWQYPLXGJIXMMV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethyl-3-methylimidazolium bromide can be used to prepare:
  • 1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide
  • 1-ethyl-3-methylimidazolium tetrafluoroborate
  • (EMI)[Cd(BTC)] (EMI = 1-ethyl-3-methylimidazolium, BTC = 1,3,5-benzenetricarboxylate), a coordination polymer

Other Notes

Hydrophobic, highly conductive imidazolium molten salt melting at ambient temp.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ionic liquid as solvent for the synthesis and crystallization of a coordination polymer:(EMI)[Cd(BTC)](EMI=1-ethyl-3-methylimidazolium,BTC=1,3,5 benzenetricarboxylate).
Liao JH, et al.
Crystal Growth & Design, 6(5), 1062-1063 (2006)
Voltammetry of oxygen in the room-temperature ionic liquids 1-ethyl-3-methylimidazolium bis ((trifluoromethyl) sulfonyl) imide and hexyltriethylammonium bis ((trifluoromethyl) sulfonyl) imide: one-electron reduction to form superoxide. Steady-state and transient behavior......
Buzzeo MC, et al.
The Journal of Physical Chemistry A, 107(42), 8872-8878 (2003)
The phase behaviour of 1-alkyl-3-methylimidazolium tetrafluoroborates; ionic liquids and ionic liquid crystals.
J. Chem. Soc., Dalton Trans., 13, 2133-2140 (1999)
Seungmin Oh et al.
The journal of physical chemistry. B, 123(39), 8274-8284 (2019-09-11)
Ionic liquids with aprotic heterocyclic anions (AHAs) have been developed for CO2 capture but have been considered for other applications as well. Previously, we have shown that AHA ILs, where the only site for reaction with CO2 is the anion
Pierre Bonhôte et al.
Inorganic chemistry, 35(5), 1168-1178 (1996-02-28)
New, hydrophobic ionic liquids with low melting points (<-30 degrees C to ambient temperature) have been synthesized and investigated, based on 1,3-dialkyl imidazolium cations and hydrophobic anions. Other imidazolium molten salts with hydrophilic anions and thus water-soluble are also described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service