Skip to Content
Merck
All Photos(1)

Key Documents

11291

Sigma-Aldrich

1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

≥97.0% (NMR)

Synonym(s):

EMIM BTI, EMIM TFSI, EMIMIm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11F6N3O4S2
CAS Number:
Molecular Weight:
391.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (NMR)

form

liquid

impurities

<0.5% water

mp

≥−15 °C (lit.)

functional group

fluoro

SMILES string

CCn1cc[n+](C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H11N2.C2F6NO4S2/c1-3-8-5-4-7(2)6-8;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

LRESCJAINPKJTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide [EMIM][N(Tf)2] is a room temperature ionic liquid (RTIL). ([EMIM][N(Tf)2]) as a non-aqueous solvent, is advantageous over traditional aprotic polar organic solvents in electrochemical investigation of electroactive species since it has low vapor pressure, high thermal stability, good conductivity and a wide electrochemical window. [EMIM][N(Tf)2] shows good solubility in CO2.

Application

[EMIM][N(Tf)2] may be used as a green solvent for the synthesis of zinc oxide (ZnO) nanoparticles under microwave conditions. It is useful in reactions which need a solvent with high CO2 solubility.

Physical form

fluorinated, hydrophobic ionic liquid

Other Notes

Enantioselective reactions in ionic liquids

Legal Information

Product protected by U.S. Patent 5,827,602 assigned to Covalent Associates, Inc. Woburn, Ma, USA

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid? liquid equilibria in binary mixtures containing substituted benzenes with ionic liquid 1-ethyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide
Shiflett, Mark B., and Anne Marie S. Niehaus
Journal of Chemical and Engineering Data, 55.1, 346-353 (2009)
Pedro Lozano et al.
Chemical communications (Cambridge, England), (7), 692-693 (2002-07-18)
Soluble Candida antarctica lipase B dissolved in ionic liquids showed good synthetic activity, enantioselectivity and operational stability in supercritical carbon dioxide for both butyl butyrate synthesis and the kinetic resolution of 1-phenylethanol processes by transesterification.
Investigation on physical and electrochemical properties of three imidazolium based ionic liquids (1-hexyl-3-methylimidazolium tetrafluoroborate, 1-ethyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide and 1-butyl-3-methylimidazolium methylsulfate)
Beigi, Ali Akbar Miran, et al.
Journal of Molecular Liquids, 177, 361-368 (2013)
Raman and ab initio study of the conformational isomerism in the 1?ethyl?3?methyl?imidazolium bis (trifluoromethanesulfonyl) imide ionic liquid
Lassegues, J. C., et al.
Journal of Raman Spectroscopy, 38.5, 551-558 (2007)
Voltammetry of oxygen in the room-temperature ionic liquids 1-ethyl-3-methylimidazolium bis ((trifluoromethyl) sulfonyl) imide and hexyltriethylammonium bis ((trifluoromethyl) sulfonyl) imide: one-electron reduction to form superoxide. Steady-state and transient behavior in the same cyclic voltammogram resulting from widely different diffusion coefficients of oxygen and superoxide
Buzzeo, Marisa C., et al.
The Journal of Physical Chemistry A, 107.42, 8872-8878 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service