Skip to Content
Merck
All Photos(3)

Documents

900771

Sigma-Aldrich

1-Ethyl-3-methylimidazolium chloride

greener alternative

>99%

Synonym(s):

1-Ethyl-3-methyl-1H-imidazolium chloride, 1-Methyl-3-ethylimidazolinium chloride, N-Methyl-N′-ethylimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClN2
CAS Number:
Molecular Weight:
146.62
Beilstein:
5163190
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.23

Quality Level

Assay

>99%

form

crystals

greener alternative product characteristics

Safer Solvents and Auxiliaries
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤2000 ppm H2O

mp

77-79 °C (lit.)

density

1.189 g/cm3

application(s)

battery manufacturing

greener alternative category

SMILES string

[Cl-].CCn1cc[n+](C)c1

InChI

1S/C6H11N2.ClH/c1-3-8-5-4-7(2)6-8;/h4-6H,3H2,1-2H3;1H/q+1;/p-1

InChI key

BMQZYMYBQZGEEY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Ionic liquids (ILs) are molten salts with melting points lower than 100 °C. They usually consist of pair of organic cation and anion. ILs exhibit unique properties such as nonvolatility, high thermal stability, and high ionic conductivity and find applications as electrolytes in lithium/sodium ion batteries and dye-sensitized solar cells. They are also used as media for synthesis of conducting polymers and intercalation electrode materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cycloaddition of carbon dioxide to butyl glycidyl ether using imidazolium
salt ionic liquid as a catalyst
Yu J, et al.
Korean Journal of Chemical Engineering, 27, 446-451 (2010)
Martin Brehm et al.
Molecules (Basel, Switzerland), 25(15) (2020-08-05)
We present 1,2,3-triazolium- and imidazolium-based ionic liquids (ILs) with aromatic anions as a new class of cellulose solvents. The two anions in our study, benzoate and salicylate, possess a lower basicity when compared to acetate and therefore should lead to
Cycloaddition of carbon dioxide to butyl glycidyl ether using imidazolium salt ionic liquid as a catalyst
Yu JI, et al.
Korean Journal of Chemical Engineering, 27, 446-451 (2010)
Ionic liquids and their solid-state analogues as materials for energy generation and storage.
MacFarlane DR, et al.
Nature Reviews. Materials, 1, 15005-15005 (2016)
Masayoshi Watanabe et al.
Chemical reviews, 117(10), 7190-7239 (2017-01-14)
Ionic liquids (ILs) are liquids consisting entirely of ions and can be further defined as molten salts having melting points lower than 100 °C. One of the most important research areas for IL utilization is undoubtedly their energy application, especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service