Skip to Content
Merck

713171

1-Ethyl-1-methylpyrrolidinium bromide

99%

Synonym(s):

N-Ethyl-N-methylpyrrolidinium bromide, [EMPyrr]Br

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H16BrN
CAS Number:
Molecular Weight:
194.11
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
418-200-5
MDL number:
Assay:
≥98.5% (T), 99%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98.5% (T), 99%

form

powder

impurities

≤1.0% water

SMILES string

[Br-].CC[N+]1(C)CCCC1

InChI

1S/C7H16N.BrH/c1-3-8(2)6-4-5-7-8;/h3-7H2,1-2H3;1H/q+1;/p-1

InChI key

KHJQQUGSPDBDRM-UHFFFAOYSA-M

General description

1-Ethyl-1-methylpyrrolidinium bromide is an ionic liquid that can be prepared by reacting N-methylpyrrolidine with bromoethane. it is conventionally used as a bromine-sequestering agent (BSA) in zinc/bromine redox flow batteries.

Application

1-Ethyl-1-methylpyrrolidinium bromide may be used in the preparation of 1-ethyl-1-methylpyrrolidinium lactate, a potential solvent for the separation of hexan-1-ene from hexane. It can also be used to prepare 1-ethyl-1-methylpyrrolidinium tribromide. It can be used as a co-catalyst during the cycloaddition of CO2 with different epoxides to form cyclic carbonates in the presence of a nona-vacant Keggin-type tricarbonyl rhenium derivative catalyst.


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Muta. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The influence of ionic liquid additives on zinc half-cell electrochemical performance in zinc/bromine flow batteries.
Rajarathnam GP, et al.
Royal Society of Chemistry Advances, 6(33), 27788-27797 (2016)
Effect of ionic liquids on the dispersion of zinc oxide and silica nanoparticles, vulcanisation behaviour and properties of NBR composites
Maciejewska, M., and M. Zaborski.
Express Polymer Letters, 8.12 (2014)
Estimation of State-of-Charge for Zinc-Bromine Flow Batteries by In Situ Raman Spectroscopy
Lee, Hyun Ju, Dong-Won Kim, and Jung Hoon Yang
Journal of the Electrochemical Society, 164.4, A754-A759 (2017)



Global Trade Item Number

SKUGTIN
713171-5G04061833338520
713171-50G04061833417560