![1-Ethyl-3-methylimidazolium bromide ≥97.0% (T)](/deepweb/assets/sigmaaldrich/product/structures/173/029/24f571b9-6ead-460b-b6cd-e3623ef49c6c/640/24f571b9-6ead-460b-b6cd-e3623ef49c6c.png)
1-Ethyl-3-methylimidazolium bromide
![1-Ethyl-3-methylimidazolium bromide ≥97.0% (T)](/deepweb/assets/sigmaaldrich/product/structures/173/029/24f571b9-6ead-460b-b6cd-e3623ef49c6c/640/24f571b9-6ead-460b-b6cd-e3623ef49c6c.png)
Empirical Formula (Hill Notation):
C6H11BrN2
CAS Number:
Molecular Weight:
191.07
Products
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing