Skip to Content
Merck
All Photos(1)

Key Documents

61609

Supelco

Dodecanoic acid

analytical standard

Synonym(s):

Lauric acid, NSC 5026

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
Beilstein:
1099477
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

plant

Quality Level

grade

analytical standard

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

43-46 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCC(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)

InChI key

POULHZVOKOAJMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Substrate for CYP 4A11

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliette Lambert et al.
Oncotarget, 5(15), 6280-6288 (2014-07-16)
We analysed the prognostic significance of minimal residual disease (MRD) level in adult patients with acute myeloid leukemia (AML) treated in the randomized gemtuzumab ozogamicin (GO) ALFA-0701 trial. Levels of WT1 and NPM1 gene transcripts were assessed using cDNA-based real-time
M Schmidt et al.
Leukemia, 28(12), 2292-2299 (2014-09-13)
To study clonal evolution in chronic myeloid leukemia (CML), we searched for BCR-ABL-independent gene mutations in both Philadelphia chromosome (Ph)-negative and Ph-positive clones in 29 chronic-phase CML patients by targeted deep sequencing of 25 genes frequently mutated in myeloid disorders.
Meng-Meng Bai et al.
Journal of agricultural and food chemistry, 63(8), 2198-2205 (2015-02-14)
Eucommia ulmoides leaves have been used as a functional food and drink in China. The purpose of this study was to identify the bioactive constituents with soluble epoxide hydrolase (sEH) inhibitory activity and anti-inflammatory properties. Twenty-seven known compounds (1-27) were
Smritilekha Bera et al.
Journal of medicinal chemistry, 51(19), 6160-6164 (2008-09-10)
Aminoglycoside antibiotics and cationic detergents constitute two classes of clinically important drugs and antiseptics. Their bacteriological and clinical efficacy, however, has decreased recently due to antibiotic resistance. We have synthesized aminoglycoside-lipid conjugates in which the aminoglycoside neomycin forms the cationic
Teigo Asai et al.
Phytochemistry, 82, 149-157 (2012-08-03)
Fourteen cyclic glycolipids, named glomerasides A-N, have been isolated from the glandular trichome exudate of Cerastium glomeratum (Caryophyllaceae). Their structures were determined by spectroscopic analysis of the glycolipids, as well as by application of the Ohrui-Akasaka method to the fatty

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service