Skip to Content
Merck
All Photos(3)

Key Documents

W283215

Sigma-Aldrich

Palmitic acid

greener alternative

natural, 98%, FG

Synonym(s):

1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
FEMA Number:
2832
Beilstein:
607489
EC Number:
Council of Europe no.:
14
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.014
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.210
FDA 21 CFR 172.860
FDA 21 CFR 173.340

vapor pressure

10 mmHg ( 210 °C)

Assay

98%

form

crystals or chunks

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

271.5 °C/100 mmHg (lit.)

mp

61-62.5 °C (lit.)

solubility

water: 0.00005 g/L at 20 °C

density

0.852 g/mL at 25 °C (lit.)

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
Pb: ≤10 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

faint; fatty

SMILES string

CCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palmitic acid is the main constituent of human milk fatty acids.[1] It is also found in sea weeds[2] and air-dried fruit of Cornus officinalis.[3]
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence that palmitic acid is absorbed assn-2 monoacylglycerol from human milk by breast-fed infants.
Innis SM, et al.
Lipids, 29(8), 541-545 (1994)
Distribution of an enzyme system producing seaweed flavor: conversion of fatty acids to long-chain aldehydes in seaweeds.
Kajiwara T, et al.
Journal of Applied Phycology, 5(2), 225-230 (1993)
Volatile flavor components of corni fructus (Cornus officinalis Sieb. Et Zucc.).
Miyazawa M & Kameoka H.
Agricultural and Biological Chemistry, 53(12), 3337-3340 (1989)
B Vessby et al.
Diabetologia, 37(10), 1044-1050 (1994-10-01)
Recent data indicate that peripheral insulin sensitivity may be influenced by dietary fat quality and skeletal muscle phospholipid fatty acid composition. During a health survey of 70-year-old men insulin sensitivity was measured by the euglycaemic hyperinsulinaemic clamp technique and the
Jonas Sieber et al.
The American journal of pathology, 183(3), 735-744 (2013-07-23)
Type 2 diabetes mellitus is characterized by dyslipidemia with elevated free fatty acids (FFAs). Loss of podocytes is a hallmark of diabetic nephropathy, and podocytes are highly susceptible to saturated FFAs but not to protective, monounsaturated FFAs. We report that

Questions

  1. What is the solubility of Palmitic Acid of the product W283215

    1 answer
    1. The product exhibits extremely low solubility in water, approximately 50 ug/L. Its solubility is as follows:
      DMF: 20 mg/ml
      DMSO: 20 mg/ml
      Ethanol: 30 mg/ml
      Ethanol:PBS (pH 7.2) (1:2): 0.25 mg/ml

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service