Skip to Content
Merck
All Photos(1)

Key Documents

55561

Supelco

Aucubin

analytical standard

Synonym(s):

Aucuboside, Rhimantin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O9
CAS Number:
Molecular Weight:
346.33
Beilstein:
50340
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C=CO[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@]1([H])C(CO)=C[C@H]2O

InChI

1S/C15H22O9/c16-4-6-3-8(18)7-1-2-22-14(10(6)7)24-15-13(21)12(20)11(19)9(5-17)23-15/h1-3,7-21H,4-5H2/t7-,8+,9+,10+,11+,12-,13+,14-,15-/m0/s1

InChI key

RJWJHRPNHPHBRN-FKVJWERZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aucubin is a chromogene iridoid glucoside isolated from the genus, Veronica L. and exhibits a broad-spectrum of biological activities such as hepatoprotective, antitoxic, neuroprotective, antiaging, antiosteoporosis, anti-inflammatory and cardioprotective effects.[1]

Application

Aucubin may be used as an analytical reference standard for the quantification of the analyte in Veronica genus using liquid chromatography coupled to mass spectrometry.[1] It may also be used as an internal standard for the determination of catalpol in biological samples using high-performance liquid chromatography coupled with atmospheric pressure chemical ionization−tandem mass spectrometry.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: plantago

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC/MS analysis of aucubin and catalpol of some Veronica species
Crisan G, et al.
Farmacia, 58, 237-247 (2010)
Antioxidant and pancreas-protective effect of aucubin on rats with streptozotocin-induced diabetes
Jin L, et al.
European Journal of Pharmacology, 582(1), 162-167 (2008)
MIC, et al.
Research Communications in Molecular Pathology and Pharmacology, 102(2), 189-204 (1998)
Yang Li et al.
Carbohydrate research, 344(16), 2270-2273 (2009-09-19)
X-ray diffraction analyses of iridoid glycoside aucubin (1) and its aglycone aucubigenin (2) are reported. It was found that crystals of 1 are orthorhombic, with P2(1)2(1)2(1) space group, both cyclopentane ring and pyran ring adopt envelope conformations, and the Glc
HPLC?PDA determination of bioactive diterpenoids from plant materials and commercial products of Andrographis paniculata
Li W and Fitzloff F.J
Journal of Liquid Chromatography and Related Technologies, 27(15), 2407-2420 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service