The half life of this product has not been determined as a product specification. However, there is a literature reference studying the stability of this compound: https://www.mdpi.com/2076-3921/10/12/1963
12247
Oleuropein
≥98.0%
Synonym(s):
(2S,3E,4S)-3-Ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-pyran-4-acetic acid 2-(3,4-dihydroxyphenyl)ethyl ester
About This Item
Recommended Products
Quality Level
Assay
≥98.0%
impurities
≤5.0% water
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
SMILES string
COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)[C@@H]1CC(=O)OCCc3ccc(O)c(O)c3
InChI
1S/C25H32O13/c1-3-13-14(9-19(29)35-7-6-12-4-5-16(27)17(28)8-12)15(23(33)34-2)11-36-24(13)38-25-22(32)21(31)20(30)18(10-26)37-25/h3-5,8,11,14,18,20-22,24-28,30-32H,6-7,9-10H2,1-2H3/b13-3+/t14-,18+,20+,21-,22+,24-,25-/m0/s1
InChI key
RFWGABANNQMHMZ-ZCHJGGQASA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- as an anti-inflammatory and antioxidant agent in hepatic steatosis on high-fat diet (HFD) fed mice[2]
- to evaluate protective effects and antioxidant properties on male reproductive function in mice[3]
- to study its inhibitory effects on bacterial (E. coli) ATP synthase[4]
- as an inhibitor of plasminogen activator inhibitor-1 (PAI-1) to test its effect on the proliferation of breast cancer cells[5]
Biochem/physiol Actions
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the half-life of this Oleuropein compound, please?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service