Skip to Content
Merck
All Photos(1)

Key Documents

41875

Supelco

trans-4-Hydroxy-L-proline

analytical standard

Synonym(s):

(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein:
81441
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

273 °C (dec.) (lit.)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1CN[C@@H](C1)C(O)=O

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1

InChI key

PMMYEEVYMWASQN-DMTCNVIQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Natural constituent of animal structural proteins such as collagen and elastin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsuan-Shu Lee et al.
Journal of gastroenterology and hepatology, 20(7), 1109-1114 (2005-06-16)
Needle liver biopsy is essential for the diagnosis of liver fibrosis and cirrhosis. However, sampling variability has been reported by semiquantitative methods. The aim of the present study was to investigate quantitatively the sampling variability of needle liver biopsy on
Ville V Meretoja et al.
Biomaterials, 34(17), 4266-4273 (2013-03-16)
In this work, we investigated the effects of lowered oxygen tension (20% and 5% O2) on the chondrogenesis and hypertrophy of articular chondrocytes (ACs), mesenchymal stem cells (MSCs) and their co-cultures with a 30:70 AC:MSC ratio. Cells were cultured for
Shahrzad Hosseininia et al.
BMC musculoskeletal disorders, 14, 18-18 (2013-01-11)
It remains to be shown whether OA shares molecular similarities between different joints in humans. This study provides evidence for similarities in cartilage molecular damage in osteoarthritic (OA) joints. Articular cartilage from osteoarthritic hip joints were analysed and compared to
Jacob Kofoed et al.
Chemical communications (Cambridge, England), (14)(14), 1482-1484 (2006-04-01)
The aldol reaction of acetone with aldehydes in aqueous medium under catalysis by zinc-proline (Zn(L-Pro)2) and secondary amines such as proline, (2S,4R)-4-hydroxyproline (Hyp) and (S)-(+)-1-(2-pyrrolidinomethyl)pyrrolidine (PMP) is shown to proceed by an enamine mechanism, as evidenced by reductive trapping of
Nis Stride et al.
American journal of physiology. Heart and circulatory physiology, 304(11), H1407-H1414 (2013-04-02)
Chronic ischemic heart disease is associated with myocardial hypoperfusion. The resulting hypoxia potentially inflicts damage upon the mitochondria, leading to a compromised energetic state. Furthermore, ischemic damage may cause excessive production of reactive oxygen species (ROS), producing mitochondrial damage, hereby

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service