Skip to Content
Merck
All Photos(1)

Key Documents

50839

Sigma-Aldrich

Catalpol

≥96% (HPLC)

Synonym(s):

(1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-hydroxy-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl-β-D-glucopyranoside, Catalpinoside, De(p-hydroxybenzoyl)catalposide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O10
CAS Number:
Molecular Weight:
362.33
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥96% (HPLC)

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](O)[C@@H]4O[C@]4(CO)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H22O10/c16-3-6-9(19)10(20)11(21)14(23-6)24-13-7-5(1-2-22-13)8(18)12-15(7,4-17)25-12/h1-2,5-14,16-21H,3-4H2/t5-,6-,7-,8+,9-,10+,11-,12+,13+,14+,15-/m1/s1

InChI key

LHDWRKICQLTVDL-PZYDOOQISA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The primary function is to stimulate the production of adrenal cortical hormones, which increases the production of sex hormones. Catalpol also exhibits anti-inflammatory activity and has shown to increase the production of androgens yielded by the adrenal gland, which can lead to increases in muscle mass.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of catalpol derivatives by characteristic neutral losses using liquid chromatography combined with electrospray ionization multistage and time-of-flight mass spectrometry.
Jun-Li Hong et al.
Rapid communications in mass spectrometry : RCM, 24(17), 2680-2686 (2010-08-27)
Cheng-Fang Wang et al.
Brain research, 1307, 158-165 (2009-10-27)
Diabetes mellitus can cause dysfunction of the central nervous system called "diabetic encephalopathy." Although insulin and various oral drugs are used to treat diabetes, they do not completely prevent the development of diabetic encephalopathy, and novel strategies for the prevention
Hui-Feng Zhu et al.
International journal of biological sciences, 6(5), 443-453 (2010-09-10)
To investigate the role and mechanism of catalpol in brain angiogenesis in a rat model of stroke, the effect of catalpol (5 mg/kg; i.p) or vehicle administered 24 hours after permanent middle cerebral artery occlusion (pMCAO) on behavior, angiogenesis, ultra-structural
Yuan-xiong Deng et al.
Journal of ethnopharmacology, 144(2), 425-432 (2012-10-06)
Huanglian Wan (HLW) is a prescription of traditional Chinese medicine (TCM), which has been used to treat diabetes mellitus for thousands of years in China. In this study we provide experimental evidence for the clinical use of HLW in the
Jing Bi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 543-550 (2012-11-21)
A large body of evidence suggests that the inflammatory reaction plays an important role in the pathogenesis of neurodegenerative diseases. Our previous studies described the neuroprotective effects of catalpol in lipopolysaccharide (LPS)-induced inflammatory models, in which catalpol was shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service