Skip to Content
Merck
All Photos(3)

Key Documents

G4796

Sigma-Aldrich

Genipin

≥98% (HPLC), powder, insulin stimulator

Synonym(s):

Methyl (1S,2R,6S)-2-hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O5
CAS Number:
Molecular Weight:
226.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Genipin, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: ≥25 mg/mL
H2O: insoluble

SMILES string

COC(=O)C1=CO[C@@H](O)C2C1CC=C2CO

InChI

1S/C11H14O5/c1-15-10(13)8-5-16-11(14)9-6(4-12)2-3-7(8)9/h2,5,7,9,11-12,14H,3-4H2,1H3/t7-,9-,11-/m1/s1

InChI key

AZKVWQKMDGGDSV-BCMRRPTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Genipin is a natural cross linking agent, which is extracted from gardenia fruit.[1] It prevents lipid peroxidation and production of nitric oxide. Genipin protects the hippocampal neurons from the toxicity of Alzheimer′s amyloid β protein. It has anti-inflammatory and anti-angiogenesis effects.[2] Genipin is involved in drug delivery system.[3]

Application

Genipin has been used:
  • in chemosensitivity assay[4]
  • in the preparation of recombinant human (rh)-odontogenic ameloblast-associated protein (ODAM) -impregnated collagen gel and in vitro mineralization assay[5]
  • in genipin gel preparation[6][7]

Biochem/physiol Actions

Genipin stimulates insulin secretion in UCP2-dependent manner (Uncoupling protein 2). Genipin is a protein, collagen, gelatin, and chitosan cross-linker.

Features and Benefits

This compound is featured on the InsR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael E Frohbergh et al.
Biomaterials, 33(36), 9167-9178 (2012-10-02)
Reconstruction of large bone defects remains problematic in orthopedic and craniofacial clinical practice. Autografts are limited in supply and are associated with donor site morbidity while other materials show poor integration with the host's own bone. This lack of integration
Effect of Sulfuric Acid Concentration on Radiological Properties of Genipin Gel Dosimeter
Al-Jarrah A, et al.
International Journal of Chemistry (Canadian Center of Science and Education), 2, 2320-4087 (2014)
UCP2 expression may represent a predictive marker of neoadjuvant chemotherapy effectiveness for locally advanced uterine cervical cancer
Imai K, et al.
Oncology Letters, 14(1), 951-957 (2017)
Ting Ting Lau et al.
Biomedical materials (Bristol, England), 7(6), 065003-065003 (2012-11-03)
Primary hepatocyte is probably the preferred cell for cell therapy in liver regeneration. However, its non-ideal proliferation capacity and rapid loss of phenotype during 2D culture compromises the quality and quantity of the transplanted hepatocytes, resulting in variable success rates
Using dairy ingredients to produce edible films and biodegradable packaging materials
Tomasula PM
Dairy-Derived Ingredients: Food and Nutraceutical Uses, 589-624 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service