Skip to Content
Merck
All Photos(1)

Documents

46725

Supelco

Salbutamol

VETRANAL®, analytical standard

Synonym(s):

α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NO3
CAS Number:
Molecular Weight:
239.31
Beilstein:
6405698
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC(C)(C)NCC(O)c1ccc(O)c(CO)c1

InChI

1S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17H,7-8H2,1-3H3

InChI key

NDAUXUAQIAJITI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706019, T706030, T706020. Salbutamol is classified under the β-agonist group of chemicals which are known to possess powerful pharmacological activities.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Salbutamol may be used as a reference standard for the determination of salbutamol in:
  • Fish plasma by solid phase extraction (SPE) followed by analysis using liquid chromatography combined with electrospray ionization triple quadrupole mass spectrometry (ESI-LC-QqQMS).
  • Feed samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) as well as supercritical fluid chromatography (SFC) with MS/MS.
  • Pig hair by HPLC in combination with ESI-MS/MS.
  • Water and swine feed by dual labeled time-resolved fluoroimmunoassay (TRFIA) using lanthanide ions-based fluorescent probes and HPLC equipped with ultraviolet (UV) detector.
  • Treated wastewater by direct injection (DI) combined with UHPLC coupled to quadrupole-linear ion trap analyser (QqLIT-MS/MS).

Biochem/physiol Actions

β2-adrenoceptor agonist

Caution

sensitive to light

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast determination of pesticides and other contaminants of emerging concern in treated wastewater using direct injection coupled to highly sensitive ultra-high performance liquid chromatography-tandem mass spectrometry.
Campos-Ma?as MC, et al.
Journal of Chromatography A, 1507(2), 84-94 (2017)
Comparison between liquid chromatography and supercritical fluid chromatography coupled to mass spectrometry for beta-agonists screening in feeding stuff.
Herpin L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1086(2), 130-137 (2018)
Simultaneous determination of ?-agonists clenbuterol and salbutamol in water and swine feed samples by dual-labeled time-resolved fluoroimmunoassay
Li M, et al.
Food Control, 73(2), 1039-1044 (2017)
Determination of ractopamine and salbutamol in pig hair by liquid chromatography tandem mass spectrometry
Chang KC, et al.
Journal of food and drug analysis, 26(2), 725-730 (2018)
Rapid screening of toxic salbutamol, ractopamine, and clenbuterol in pork sample by high-performance liquid chromatography?UV method.
Yan K, et al.
Journal of food and drug analysis, 24(2), 277-283 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service