
Salbutamol

Synonym(s):
α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol
Empirical Formula (Hill Notation):
C13H21NO3
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique