Skip to Content
Merck
All Photos(1)

Key Documents

32379

Supelco

Zilpaterol hydrochloride

VETRANAL®, analytical standard

Synonym(s):

(6RS,7RS)-4,5,6, 7-Tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]imidazo[ 4,5,1-jk][1]benzazepin-2(1H)-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19N3O2 · HCl
CAS Number:
Molecular Weight:
297.78
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.[n]21c3c(nc2O)cccc3[C@H]([C@@H](CC1)NC(C)C)O

InChI

1S/C14H19N3O2.ClH/c1-8(2)15-11-6-7-17-12-9(13(11)18)4-3-5-10(12)16-14(17)19;/h3-5,8,11,13,15,18H,6-7H2,1-2H3,(H,16,19);1H/t11-,13-;/m1./s1

InChI key

GIEFXLLRTJNFGT-LOCPCMAASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zilpaterol belongs to the group of β-adrenergic agonists, used in the treatment of lung diseases due to its brochodilating nature. It is also widely used in veterinary medicine as a growth promoter for increasing muscle gain and decreasing fat.

Application

Zilpaterol hydrochloride analytical standard can be used as follows:
  • Determination of zilpaterol in cattle plasma and tissue samples using solid phase extraction (SPE) well plate of 96 samples for sample preparation and subsequent analysis by liquid chromatography-triple quadrupole mass spectrometry (LC-MS/MS)
  • Multi-residue analysis of 3 β-agonists, zilpaterol, clenbuterol, and ractopamine in a variety of meat samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • QuEChERS-based preparation of two types of animal tissue samples, muscle and viscera, for the analysis of seven β-agonists by liquid chromatography/tandem mass spectrometry
  • Development and validation of an LC-MS/MS method to estimate zilpaterol, ractopamine, and clenbuterol in bovine tissue samples
  • Simultaneous quantitative determination of spectinomycin, halquinol, zilpaterol, and melamine in animal feed samples by LC-MS/MS

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Rivera-Villegas et al.
Asian-Australasian journal of animal sciences, 32(2), 209-216 (2018-07-31)
We compare the effects of three different approved sources of supplemental zilpaterol on growth-performance responses and carcass characteristics of finishing lambs. Twenty four Pelibuey×Katahdin lambs (46.75±2.43 kg) were used in a 33-day feeding trial. Lambs were fed a dry rolled
J A Hagenmaier et al.
Journal of animal science, 94(12), 5064-5076 (2017-01-04)
Feedlot cattle ( = 1,395; BW = 568 ± 43 kg) were used to evaluate the effects of shade on animal welfare, growth performance, and carcass quality during the summer of 2013 in a Kansas commercial feedlot. Seven lots of
Ennet Moholisa et al.
Journal of the science of food and agriculture, 97(8), 2375-2381 (2016-10-08)
Animal age as determined by number of permanent incisors (p. i.) is used in classification of beef carcasses to describe expected meat tenderness. However, animals differing in age are reared under different production systems (pasture or feedlot). In addition to
Natalia Cernicchiaro et al.
Foodborne pathogens and disease, 13(9), 517-525 (2016-06-16)
The efficacy of a Salmonella vaccine for reducing fecal shedding of Salmonella during the finishing period and lymph node (LN) carriage at harvest was investigated in commercial feedlot cattle. The study was designed as a pen-level randomized complete block with
P J Kononoff et al.
Journal of animal science, 95(1), 194-200 (2017-02-09)
A total of 2,948 steers (mean initial BW = 568.9 ± 49.4 kg) were used to evaluate the effect of the LEP R25C SNP genotype on feed intake, growth performance, and carcass characteristics over time. Steers were grouped into 5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service