Skip to Content
Merck
All Photos(1)

Key Documents

C5423

Sigma-Aldrich

Clenbuterol hydrochloride

≥95% (TLC), powder, β2-adrenoceptor agonist

Synonym(s):

4-Amino-α-(t-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Cl2N2O · HCl
CAS Number:
Molecular Weight:
313.65
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Clenbuterol hydrochloride, ≥95%

Assay

≥95%

form

powder

drug control

Home Office Schedule 4.2

storage temp.

2-8°C

SMILES string

Cl.CC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1

InChI

1S/C12H18Cl2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H

InChI key

OPXKTCUYRHXSBK-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

Application

Clenbuterol hydrochloride has been used:
  • as an agonist for β-adrenergic receptor (β-AR) from chick skeletal muscle
  • to induce hypertrophic effects in masseter, digastric, and temporalis muscles of rat
  • as a reference β-agonist standard in tandem mass spectrometry(MS-MS)

Biochem/physiol Actions

Clenbuterol hydrochloride is a β2-adrenoceptor agonist and bronchodilator. It is effective in improving peak expiratory flow rate (PEFR) in asthma patients. Clenbuterol is also a performance-enhancing drug as it stimulates the central nervous system and improves oxygen transport. Clenbuterol elicits cardio protective functionality in patients with dilated cardiomyopathy.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelka Pleadin et al.
Meat science, 86(3), 733-737 (2010-07-30)
The aim of this study was to determine the level of clenbuterol residues in muscle tissue of pigs after repeat administration in a growth-promoting dose. An anabolic dose of clenbuterol (20 μg/kg body mass per day) was administered orally to
I Semkova et al.
Brain research. Brain research reviews, 30(2), 176-188 (1999-10-19)
Neurotrophins and other neurotrophic factors have been shown to support the survival and differentiation of many neuronal populations of the central and peripheral nervous system. Therefore, administering neurotrophic factors could represent an alternative strategy for the treatment of acute and
A trial of clenbuterol in bronchial asthma.
Anderson G and Wilkins E
Thorax, 32(6), 717-719 (1977)
C A Izeboud et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 48(9), 497-502 (1999-10-16)
To investigate the suppressive effects of the beta-agonist clenbuterol on the release of TNF-alpha and IL-6 in a lipopolysaccharide (LPS)-model of inflammation, both in vitro and in vivo. Human U-937 cell line (monocyte-derived macrophages), and male Wistar rats (200-250 g).
Clinton E Canal et al.
Learning & memory (Cold Spring Harbor, N.Y.), 15(9), 677-686 (2008-09-06)
Infusions of CREB antisense into the amygdala prior to training impair memory for aversive tasks, suggesting that the antisense may interfere with CRE-mediated gene transcription and protein synthesis important for the formation of new memories within the amygdala. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service