HT1079
Weigert′s Iron Hematoxylin Set
Set
Synonym(s):
Weigert’s iron hematoxylin solution
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
solution
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
application(s)
hematology
histology
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
Application
Recommended in lengthy or acidic stain procedures. Resists decolorization better than aluminum based hematoxylin formulations.
Components
Weigert′s iron hematoxylin solution set consists of HT107 Part A and HT109 Part B.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1
Storage Class Code
3 - Flammable liquids
Flash Point(F)
60.0 °F
Flash Point(C)
15.55 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 11(1), e0144523-e0144523 (2016-01-20)
Single-vessel, intracoronary infusion of stem cells under stop-flow conditions has proven safe but achieves only limited myocardial coverage. Continuous flow intracoronary delivery to one or more coronary vessels may achieve broader coverage for treating cardiomyopathy, but has not been investigated.
Arteriosclerosis, thrombosis, and vascular biology, 40(6), 1479-1490 (2020-04-10)
Enhanced expression of PAI-1 (plasminogen activator inhibitor-1) has been implicated in atherosclerosis formation in humans with obesity and metabolic syndrome. However, little is known about the effects of pharmacological targeting of PAI-1 on atherogenesis. This study examined the effects of
Stem cells translational medicine, 6(2), 394-404 (2017-02-14)
Benign prostatic hyperplasia (BPH) is the overgrowth of prostate tissues with high prevalence in older men. BPH pathogenesis is not completely understood, but it is believed to be a result of de novo overgrowth of prostatic stroma. In this study
Journal of molecular endocrinology, 64(2), 77-90 (2019-12-20)
Lack of leptin production in ob/ob mice results in obesity and prediabetes that could be partly reversed by leptin supplementation. In the hypothalamus, leptin supports the production of prolactin-releasing peptide (PrRP), an anorexigenic neuropeptide synthesized and active in the brain.
Frontiers in molecular neuroscience, 10, 337-337 (2017-11-09)
Collagen VI is an extracellular matrix (ECM) protein playing a key role in skeletal muscles and whose deficiency leads to connective tissue diseases in humans and in animal models. However, most studies have been focused on skeletal muscle features. We
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service