Skip to Content
Merck
All Photos(1)

Key Documents

197378

Sigma-Aldrich

Picric acid

moistened with water, ≥98%

Synonym(s):

2,4,6-Trinitrophenol, PA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Colour Index Number:
10305
Beilstein:
423400
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

7.9 (vs air)

vapor pressure

1 mmHg ( 195 °C)

Assay

≥98%

form

powder or crystals (with a top layer of Liquid)

contains

≥35% water

expl. lim.

0.01 %

concentration

≥50-<70%

mp

122-123 °C (dried material) (lit.)

solubility

alcohol: 1 (g/12 mL)(lit.)
benzene: 1 g/10 mL
chloroform: 1 (g/35 mL)
diethyl ether: 1 (g/65 mL)

density

1.763 g/cm3

functional group

nitro

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Picric acid (PA, 2,4,6-trinitrophenol) is a polynitrated aromatic acid. It is strongly acidic in nature. Since it readily dissolves in water, it can easily pollute the environment on exposure. Sensitive detection of PA by tris-imidazolium salt, fluorescent [3+2] self-assembled nanoscopic organic cage and rhodamine based sensors have been reported. It is widely used for the determination of various organic compounds by spectrophotometric and extractive spectrophotometric methods.

Application

Picric acid has been used in the preparation of Bouin′s fixative and picric acid/sodium azide/multi-walled carbon nanotubes.
Picric acid was used in the quantitative determination of bisoprolol.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Desen. Expl. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bijan Roy et al.
The Journal of organic chemistry, 78(3), 1306-1310 (2013-01-12)
Two new anthracene-functionalized fluorescent tris-imidazolium salts have been synthesized, characterized, and proven to be selective sensors for picric acid, which is a common constituent of many powerful explosives. Theoretical studies revealed an unusual ground-state electron transfer from picrate anion to
Mario Sanhueza et al.
Journal of visualized experiments : JoVE, (111)(111), doi:10-doi:10 (2016-05-24)
Most studies on morphogenesis rely on qualitative descriptions of how anatomical traits are affected by the disruption of specific genes and genetic pathways. Quantitative descriptions are rarely performed, although genetic manipulations produce a range of phenotypic effects and variations are
Alina-Diana Panainte et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 117(2), 520-524 (2013-12-18)
Bisoprolol is a drug belonging to beta blockers drugs used primarily for the treatment of cardiovascular diseases. A spectrophotometric method for quantitative determination of bisoprolol was developed based on the formation of a complex combination between bisoprolol and picric acid.
Koushik Acharyya et al.
Chemical communications (Cambridge, England), 50(99), 15788-15791 (2014-11-06)
Dynamic covalent imine chemistry has been utilized to synthesize a fluorescent [3+2] self-assembled nanoscopic organic cage. The fluorescent nature of the reduced analogue of the cage was further exploited for the highly selective detection of the explosive picric acid (PA).
Rhodamine based selective turn-on sensing of picric acid.
Sivaraman G, et al.
Royal Society of Chemistry Advances, 4(58), 30828-30831 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service