Skip to Content
Merck
All Photos(3)

Key Documents

F7252

Sigma-Aldrich

Fast Green FCF

Dye content ≥85 %, powder

Synonym(s):

Food green 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C37H34N2O10S3Na2
CAS Number:
Molecular Weight:
808.85
Colour Index Number:
42053
Beilstein:
5718212
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Fast Green FCF, Dye content ≥85 %

form

powder

Quality Level

composition

Dye content, ≥85%

mp

290 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL, clear

SMILES string

[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)C(=C3/C=CC(\C=C3)=[N+](\CC)Cc4cccc(c4)S([O-])(=O)=O)\c5ccc(O)cc5S([O-])(=O)=O

InChI

1S/C37H36N2O10S3.2Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);;/q;2*+1/p-2

InChI key

RZSYLLSAWYUBPE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fast Green FCF is one of the triarylmethane dyes, that is biocompatible. This triphenylmethane color additive is mainly used in coloring food, drugs and cosmetics.

Application

Fast Green FCF dye is used for protein staining in IEF, native PAGE and SDS-PAGE. Fast Green staining is linear over a wider range of protein concentrations than Brilliant Blue R. After electrophoresis, fixing the proteins in the gel is recommended for maximum sensitivity. The gel can then be stained for 2 hr with 0.1% Fast Green FCF in either 30% (v/v) ethanol, 10% (v/v) acetic acid or in 7% acetic acid. The gel can be destained with either 30% (v/v) ethanol, 10% (v/v) acetic acid, or 7% acetic acid. The stained gel can be scanned at 625 nm. The detection sensitivity is approx. 30% that with Brilliant Blue R.
Fast Green FCF has been used for in staining of menisci from rabbit and of osteochondral tissues.

Biochem/physiol Actions

Fast Green FCF can be considered as a safe stain to detect tumor. In recent years, this has gained importance as an important drug to treat amyloid-deposit diseases.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Food Colors
Corradini MG
Encyclopedia of Agriculture and Food Systems, 291-296 (2019)
Sarah Zerimech et al.
Neuropharmacology, 166, 107951-107951 (2020-01-17)
Cortical spreading depression (CSD) is a wave of transient network hyperexcitability leading to long lasting depolarization and block of firing, which initiates focally and slowly propagates in the cerebral cortex. It causes migraine aura and it has been implicated in
Chitosan-glycerol phosphate/blood implants increase cell recruitment, transient vascularization and subchondral bone remodeling in drilled cartilage defects
Chevrier A, et al.
Osteoarthritis and Cartilage, 15(3), 316-327 (2007)
Fast green FCF alleviates pain hypersensitivity and down-regulates the levels of spinal P2X4 expression and pro-inflammatory cytokines in a rodent inflammatory pain model
Xu F, et al.
Frontiers in Pharmacology, 9, 534-534 (2018)
Amanda M Hamilton et al.
PloS one, 14(12), e0214107-e0214107 (2019-12-04)
Osteoarthritis (OA) is characterized by cartilage degradation and chronic joint inflammation. Mesenchymal stem cells (MSCs) have shown promising results in OA, but their mechanism of action is not fully understood. We hypothesize that MSCs polarize macrophages, which are strongly associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service