Skip to Content
Merck
All Photos(3)

Key Documents

04693159001

Roche

cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail

Tablets provided in EASYpacks

Synonym(s):

Protease Inhibitor Tablets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21
form:
tablet
solubility:
aqueous buffer: soluble
water: soluble
storage temp.:
2-8°C

form

tablet

packaging

pkg of 30 tablets (individually packed in foil blister packs)

manufacturer/tradename

Roche

concentration

(Starting concentration: 1 tablet contains protease inhibitors sufficient for a 10 ml cell extract.)

technique(s)

inhibition assay: suitable

solubility

aqueous buffer: soluble
water: soluble

absorption

0.08 at 280 nm

storage temp.

2-8°C

General description

cOmplete, Mini, EDTA-free tablets, inhibit a broad spectrum of serine and cysteine proteases. In contrast to other cOmplete tablets they do not contain EDTA, leaving the stability and the function of metal-dependant proteins unaffected. The affinity purification of Poly-His-tagged fusion proteins using IMAC (Immobilized Metal Affinity Chromatography) is also facilitated (no dialysis necessary).
Due to the optimized composition of the tablets they show excellent inhibition of serine and cysteine proteases, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria. cOmplete, Mini, EDTA-free tablets, contains both irreversible and reversible protease inhibitors. Metallo- and aspartic proteases are not inhibited.

cOmplete Mini, EDTA-free Tablets are identical to cOmplete Mini Tablets, the only difference being that no EDTA or other chelating (e.g. EGTA) agents are included.

Specificity

Mixture of several protease inhibitors. For the inhibition of serine, cysteine, but not metalloproteases.

Application

cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail has been used as a component of
  • nonidet P-40 (NP-40) buffer for the preparation of cell lysate of neuroblastoma cells
  • tissue lysis buffer for homogenisation and lysis of prefrontal cortex tissues
  • ice-cold radioimmunoprecipitation assay (RIPA) buffer for the lysis of Chinese hamster ovary (CHO-K1) cells

cOmplete, Mini, EDTA-free, is used for the inhibition of serine and cysteine proteases in bacterial, yeast, plant, and animal cell extracts.
cOmplete, Mini, EDTA-free Tablets, are used for the inhibition of proteolytic activity in small volumes (up to 10 ml) in which EDTA may interfere with protein stability (e.g., metal-containing proteins) or subsequent assays. Since EDTA interferes with IMAC (Immobilized Metal Affinity Chromatography), cOmplete, Mini, EDTA-free, is preferentially used in the isolation process of Poly-His-tagged fusion proteins.
If it is necessary to inhibit proteolytic activity in a large volume (up to 50 ml), we recommend to use cOmplete, EDTA-free.

Features and Benefits

  • Convenience: Simply push the quick-dissolving tablet through the foil packaging of the EASYpack into your buffer.
  • Complete protection: Instantly protect your proteins against a broad range of proteases (see Table 1).
  • Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.
  • Safe: Choose non-toxic inhibitors that pose no risk to you or those around you.

Packaging

30 individually packed cOmplete Protease Inhibitor Cocktail Tablets in foil blisters. Each tablet is sufficient for a volume of 10 ml solution.

Quality

Performance-tested with pancreas extract.

Physical form

Tablets in foil blister packs

Preparation Note

Working concentration: 1 tablet per 10 ml extraction solution
Working solution: Handling

Carefully push the tablet through the foil packaging using the base of your thumb (not fingernail) to prevent the breakage of tablets.

Preparation of Working Solution

One cOmplete Mini EDTA-free tablet is sufficient for the inhibition of the proteolytic activity in 10 ml extraction solution. When very high proteolytic activity is present, one tablet should be used for 7 ml extraction buffer. Tablets can be added directly to the extraction medium. Alternatively a stock solution (7 X conc.) can be prepared.

Stock solution (7 X conc.)

Dissolve one cOmplete Mini EDTA-free tablet in 1.5 ml dist. water or in 1.5 ml 100 mM phosphate buffer, pH 7.0.
Storage conditions (working solution): The stock solution can be stored at 2 to 8 °C for 1 to 2 weeks, or at least 12 weeks at -15 to -25 °C.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 7x stock solutions in 1.5 ml water or 100 mM phosphate buffer, pH 7.0. Stock solution is stable for 1-2 weeks at 2 to 8 °C, or at least 12 weeks at -15 to -25 °C. Can be used in thiol-containing solutions at 15 to 25 °C.

Storage and Stability

Store at 2 to 8 °C. (storage at -15 to -25 °C is also possible)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

cOmplete is a trademark of Roche

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katy Stokes et al.
Nucleic acids research, 48(14), 8128-8145 (2020-06-26)
The eukaryotic replisome must faithfully replicate DNA and cope with replication fork blocks and stalling, while simultaneously promoting sister chromatid cohesion. Ctf18-RFC is an alternative PCNA loader that links all these processes together by an unknown mechanism. Here, we use
David S Guttery et al.
Life science alliance, 3(12) (2020-10-28)
Cells use fatty acids (FAs) for membrane biosynthesis, energy storage, and the generation of signaling molecules. 3-hydroxyacyl-CoA dehydratase-DEH-is a key component of very long chain fatty acid synthesis. Here, we further characterized in-depth the location and function of DEH, applying
Comparative Phosphoproteomic Profiling of Type III Adenylyl Cyclase Knockout and Control, Male, and Female Mice
Zhou Y, et al.
Frontiers in Cellular Neuroscience, 13, 34-34 (2019)
Fang Tian et al.
Drug design, development and therapy, 14, 1041-1055 (2020-03-27)
Many studies have confirmed that high myopia is related to the high prevalence of cataracts, which results from apoptosis of lens epithelial cells (LECs) due to endoplasmic reticulum stress. Krüppel-like factor 6 (KLF6) is a tumor suppressor that is involved
Huimin Dong et al.
Oncology reports, 45(2), 595-605 (2021-01-09)
Rho family GTPase 3 (RND3) is involved in multiple physiological activities involving the Rho kinase‑dependent signaling pathway. The present study revealed a novel role of RND3 in the regulation of apoptosis in the brain. Using immunofluorescence and TUNEL assays, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service