Skip to Content
Merck
All Photos(4)

Key Documents

COUEDTAF-RO

Roche

cOmplete ULTRA Tablets, EDTA-free, glass vials Protease Inhibitor Cocktail

Tablets provided in glass vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21
form:
tablet
solubility:
aqueous buffer: soluble
storage temp.:
2-8°C

form

tablet

usage

sufficient for 50 mL (05892953001)
sufficient for 50 mL (06538282001)

packaging

pkg of 2 × 10 tablets (05892953001)
pkg of 6 × 10 tablets (06538282001)

manufacturer/tradename

Roche

solubility

aqueous buffer: soluble

absorption

0.4 at 280 nm (1x solution)

storage temp.

2-8°C

General description

cOmplete ULTRA Tablets, EDTA-free contain the most powerful protease inhibitor cocktail for the optimal protection of your proteins against protease degradation. One cOmplete ULTRA Tablet, EDTA-free is sufficient for a volume of 50 ml extraction solution. cOmplete ULTRA Tablets contain both irreversible and reversible protease inhibitors. Tablets are supplied in glass vials. Alternatively, these tablets are also available in EASYpack blister packs. EASYpacks provide the most convenient form of delivery - single tablets are easily pushed through the blister foil directly into the buffer.


cOmplete ULTRA Tablets EDTA-free are identical to cOmplete ULTRA Tablets, the only difference being that no EDTA or other chelating agents (e.g. EGTA) are included.


Contents
Tablets in a glass vial. Each tablet is sufficient for a volume of 50ml extraction solution.

Specificity

Inhibits serine proteases, cysteine proteases. Does not inhibit metalloproteases.

Application

During cell lysis and purification, target proteins are at risk of being cleaved by proteases, which are naturally present in cells. To ensure that full-length proteins are obtained, cOmplete ULTRA Tablets are used for the inhibition of an extended range of proteases. This new formulation shows high inhibition ratios due to its broadened range of inhibitors compared to standard cOmplete Tablets.
This new protease inhibitor cocktail now inhibits aspartic proteases, as well as serine and cysteine proteases. cOmplete ULTRA Tablets were developed for improved efficiency on different cell types. Best performance is guaranteed in lysates of bacteria, yeast, insect cells, and a wide range of higher cell lines.
This tablet does not contain EDTA; therefore metalloproteases are not inhibited.
cOmplete ULTRA Tablets, EDTA-free, glass vials Protease Inhibitor Cocktail has been used as a component of
  • ice-cold radioimmunoprecipitation assay buffer (RIPA) for the lysis of retinal ganglion cells
  • lysis buffer for green fluorescent protein (GFP)- cell-penetrating peptides (CPPs) fusion protein isolation from E coli
  • homogenization buffer for mice protein isolation from intestinal tissue

Features and Benefits

  • Protect your precious proteins.
Choose cOmplete ULTRA for even more efficient protease inhibition in eukaryotic, yeast, and bacterial cell lysates.
  • Why settle for less.
Benefit from 7 powerful protease inhibitors – more than in other EDTA-free inhibitor tablets.
  • Defend against a broad range of proteases.
Reliably inhibit serine, cysteine, and aspartic proteases.
  • Convenient and safe tablet format.
Eliminate weighing small amounts of individual inhibitors which might be hazardous. Simply drop a tablet
into your buffers.

Components

Proprietary mixture of several protease inhibitors with broad inhibitory specificity.

Physical form

Tablets in a glass vial. Each tablet is sufficient for a volume of 50 ml extraction solution.

Preparation Note

Working concentration: 1 Tablet per 50 ml extraction solution.
Working solution: Handling

Carefully push the tablet through the foil packaging using the base of your thumb (not your fingernail) to prevent the breakage of tablets.

Preparation of Working Solution

One tablet is sufficient for the inhibition of proteolytic activity in 50 ml extraction solution. If very high proteolytic activity is present, 2-3 tablets should be used for 50 ml extraction buffer. The tablets can be added directly to the extraction medium.

Stock Solution (2x conc.)

Alternatively to the direct use with lysates, a 2x stock solution can be prepared, with the following procedure:
• Add one cOmplete ULTRA Tablet, EDTA-free to 25 ml double-dis­tilled water in a PE or PP tube.
• Vortex 4 min, until the tablet is dissolved.
• The stock solution is ready to use even if there are very fine parti­cles still visible in solution. These particles do not interfere with the inhibition performance of the tablets.
• This stock solution is stable for 4 weeks, stored at 2 to 8 °C, and for at least 4 weeks at  -15 to  -25 °C as well.
• During storage of the 2x stock solution, the particles can sediment. Briefly vortex the stock solution prior to use.

Storage conditions (working solution): The stock solution in water is stable for 4 weeks, stored at 2 to 8 °C, and for at least 4 weeks at  -15 to -25 °C as well.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. For the stock solution, use water only.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

cOmplete is a trademark of Roche

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cell-penetrating peptide sequence and modification dependent uptake and subcellular distribution of green florescent protein in different cell lines
Patel SG, et al.
Scientific Reports, 9(1), 6298-6298 (2019)
George E Gentsch et al.
Cold Spring Harbor protocols, 2014(5), doi:10-doi:10 (2014-05-03)
Chromatin immunoprecipitation (ChIP) combined with genomic analysis techniques provide a global snapshot of protein-DNA interactions in the context of chromatin, yielding insights into which genomic loci might be regulated by the DNA-associated protein under investigation. This protocol describes how to
Strain specific maturation of Dendritic cells and production of IL-1beta controls CD40-driven colitis
Wagner AO, et al.
PLoS ONE, 14(1), e0210998-e0210998 (2019)
Ezh2 does not mediate retinal ganglion cell homeostasis or their susceptibility to injury
Cheng L, et al.
Testing, 13(2), e0191853-e0191853 (2018)
Barbara Mosca et al.
Nature communications, 4, 1541-1541 (2013-02-28)
Muscle strength declines with age in part due to a decline of Ca(2+) release from sarcoplasmic reticulum calcium stores. Skeletal muscle dihydropyridine receptors (Ca(v)1.1) initiate muscle contraction by activating ryanodine receptors in the sarcoplasmic reticulum. Ca(v)1.1 channel activity is enhanced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service