Skip to Content
Merck
All Photos(2)

Key Documents

S8830

Sigma-Aldrich

Protease Inhibitor Cocktail

SIGMA FAST, EDTA-Free, tablet, for the inhibition of serine, cysteine, aspartic and metalloproteases, for use in purification of Histidine-tagged proteins

Synonym(s):

Protease Inhibitor Cocktail

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
form:
tablet
storage temp.:
2-8°C

Product Name

SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins

biological source

bovine lung

Quality Level

form

tablet

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free is a water-soluble tablet formulation designed to inhibit serine, cysteine, aspartic and metalloproteases. The tablets contain no strong metal chelators, such as EDTA, and are compatible with sample processing by immobilized metal affinity chromatography (IMAC). The tablets also contain a unique, proprietary formulation that includes a pepstatin A salt for additional protection.

Specificity

Inhibits serine, cysteine, aspartic, and metalloproteases.

Application

The broad specificity of SIGMAFAST Protease Inhibitor Cocktail Tablets makes them suitable for use in the purification of histidine-tagged proteins and as a general-purpose protease inhibitor cocktail. The tablets can also be used as components of buffers for cell lysis, and have been optimized and tested for use with HIS-Select® media.

Features and Benefits

EDTA-Free: Contains no strong metal chelators, such as EDTA, which might interfere with IMAC processing.

Pepstatin A: Contains a unique, proprietary formulation that includes a pepstatin A salt for additional protection not normally obtained in a tablet format.

Broad Specificity: Inhibits serine, cysteine, aspartic, and metalloproteases, providing broad protection for protein samples.

High Yield: One tablet makes 100 mL of cocktail, and is recommended for the inhibition of the proteases present in a maximum of 20 g of cell mass in 100 mL of extract.

Long Stability: The tablets are stable as supplied for at least 2 years at 2–8 °C, and the reconstituted protease inhibitor solution (1´ or 10´) is stable for at least 2 weeks at 2–8 °C.

Components

AEBSF
Bestatin
E-64
Pepstatin A
Phosphoramidon
Leupeptin
Aprotinin

Quantity

One tablet makes 100 mL of cocktail. One tablet is recommended for the inhibition of the proteases present in a maximum of 20 g of cell mass in 100 mL of extract.

Physical form

Tablet that is water soluble

Storage and Stability

The tablets are stable as supplied for at least 2 years at 2–8 °C. The reconstituted protease inhibitor solution (1´ or 10´) is stable for at least 2 weeks at 2–8 °C.

Legal Information

HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany
SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin M Nitsche et al.
BMC genomics, 13, 380-380 (2012-08-10)
Filamentous fungi are confronted with changes and limitations of their carbon source during growth in their natural habitats and during industrial applications. To survive life-threatening starvation conditions, carbon from endogenous resources becomes mobilized to fuel maintenance and self-propagation. Key to
Zilun Wei et al.
Life sciences, 232, 116632-116632 (2019-07-07)
The inflammation modulation effects of mesenchymal stromal cell-derived exosomes (MSC-EXO) are well established. We aimed to explore the mechanism behind the inflammatory responses of numerous exosomal cargo molecules that have been neglected in molecular biology research, and to develop an
Stephan G Mauracher et al.
Phytochemistry, 99, 14-25 (2014-01-28)
Tyrosinases catalyze two initial reaction steps in the formation of melanin. Purification of tyrosinases had always been a process accompanied with various problems caused by enzymatic browning processes. Here, an approach is presented for the purification of the latent enzyme
The effect of pegylated granulocyte colony-stimulating factor treatment prior to experimental mastitis in lactating Holsteins
Powell EJ, et al.
Dairy science & technology (2018)
Katarzyna Rybak et al.
Developmental cell, 29(5), 607-620 (2014-06-03)
Plant cytokinesis is initiated in a transient membrane compartment, the cell plate, and completed by a process of maturation during which the cell plate becomes a cross wall. How the transition from juvenile to adult stages occurs is poorly understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service