Skip to Content
Merck
All Photos(1)

Key Documents

59678

Supelco

Potassium phosphate dibasic

suitable for HPLC, LiChropur, 99.0-101.0% (T)

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
EC Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21
Pricing and availability is not currently available.

Assay

99.0-101.0% (T)

form

powder or crystals

quality

LiChropur

technique(s)

HPLC: suitable

loss

≤1.0% loss on drying

pH

8.7-9.3

solubility

H2O: 5%, clear, colorless

UV absorption

λ: 230 nm Amax: ≤0.15
λ: 240 nm Amax: ≤0.12
λ: 250 nm Amax: ≤0.11
λ: 310 nm Amax: ≤0.05
λ: 500 nm Amax: ≤0.02

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Development of Simultaneous Analytical Method for Imidazolinone Herbicides from Livestock Products by UHPLC-MSMS.: Potassium phosphate dibasic is utilized in the development of a UHPLC-MSMS method for the simultaneous analysis of imidazolinone herbicides in livestock products, enhancing analytical accuracy and efficiency (Heo et al., 2022).

  • High-Throughput Method for Natural Organic Matter Hydrophobicity Assessment Using an Ionic Liquid-Based Aqueous Two-Phase System.: This research employs potassium phosphate dibasic in an aqueous two-phase system for assessing the hydrophobicity of natural organic matter, contributing to environmental science methodologies (Wang et al., 2021).

  • Using aquatic vegetation to remediate nitrate, ammonium, and soluble reactive phosphorus in simulated runoff.: The study discusses the use of potassium phosphate dibasic in conjunction with aquatic vegetation to remediate nutrient pollution in runoff, highlighting its environmental applications (Moore et al., 2016).

  • β-lactoglobulin stabilized nanemulsions--Formulation and process factors affecting droplet size and nanoemulsion stability.: This article investigates the role of potassium phosphate dibasic in stabilizing β-lactoglobulin nanemulsions, emphasizing its impact on formulation and stability in pharmaceutical applications (Ali et al., 2016).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley Maynard et al.
Cell, 182(5), 1232-1251 (2020-08-22)
Lung cancer, the leading cause of cancer mortality, exhibits heterogeneity that enables adaptability, limits therapeutic success, and remains incompletely understood. Single-cell RNA sequencing (scRNA-seq) of metastatic lung cancer was performed using 49 clinical biopsies obtained from 30 patients before and

Questions

  1. What is the sulfate concentration in this product?

    1 answer
    1. Sulfate concentration is not a tested parameter for this product and therefore is not assigned a specification.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service