79762
Ammonium phosphate dibasic
suitable for HPLC, LiChropur™, 99.0-101.0% (T)
Synonym(s):
Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)
About This Item
Recommended Products
Quality Level
Assay
99.0-101.0% (T)
form
crystals
quality
LiChropur™
technique(s)
HPLC: suitable
mp
155 °C (dec.) (lit.)
solubility
H2O: 50 mg/mL, clear, colorless
UV absorption
λ: 260 nm Amax: ≤0.04
λ: 270 nm Amax: ≤0.04
λ: 280 nm Amax: ≤0.03
λ: 290 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02
SMILES string
N.N.OP(O)(O)=O
InChI
1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)
InChI key
MNNHAPBLZZVQHP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Human Bone Paleoproteomics Utilizing the Single-Pot, Solid-Phase-Enhanced Sample Preparation Method to Maximize Detected Proteins and Reduce Humics.: This study uses ammonium phosphate dibasic in the preparation process to improve protein detection and reduce contaminants, enhancing the accuracy and efficiency of paleoproteomics analyses (Cleland TP, 2018).
- Effect of Mg(2+) doping on beta-alpha phase transition in tricalcium phosphate (TCP) bioceramics.: Ammonium phosphate dibasic plays a crucial role in the synthesis and phase transition of bioceramics, contributing to advancements in biomaterials and medical implants (Frasnelli M, Sglavo VM, 2016).
- Development and validation of a stability-indicating HPLC method for determination of voriconazole and its related substances.: This research employs ammonium phosphate dibasic in the mobile phase for HPLC analysis, ensuring precise and reliable quantification of pharmaceutical compounds and their impurities (Gu P, Li Y, 2009).
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service