Skip to Content
Merck
All Photos(1)

Key Documents

407709

Sigma-Aldrich

Insulin, Zinc, Human, Recombinant, P. pastoris

Synonym(s):

Insulin, Zinc, Human, Recombinant, P. pastoris

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
biological source:
yeast (Pichia pastoris)
form:
solid
Assay:
≥95—105%
Pricing and availability is not currently available.

biological source

yeast (Pichia pastoris)

Quality Level

sterility

non-sterile

Assay

≥95—105%

form

solid

specific activity

≥25 IU/mg (dry basis)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<10 EU/mg Endotoxin (EU/mg protein)

color

white to off-white

solubility

0.01 M HCl: soluble

shipped in

wet ice

storage temp.

−20°C

InChI key

PBGKTOXHQIOBKM-FHFVDXKLSA-N

General description

Recombinant, human insulin (zinc) expressed in P. pastoris. A peptide anabolic hormone produced by pancreatic β-cells.
Recombinant, human insulin (zinc) expressed in P. pastoris. Peptide anabolic hormone produced by pancreatic β-cells. Increases the synthesis of protein, fat, glycogen, and substrate oxidation in mitochondria. Interacts with membrane insulin receptor to cause its biological actions.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Insulin is practically insoluble from pH 4.5-7.0.

Reconstitution

Following reconstitution store in the refrigerator (4°C). Stock solutions are stable for up to 6 weeks at 4°C.

Other Notes

Mohan, C., et al. 1989. Curr. Topics Cell. Reg.30, 105.
Johnson, I.S. 1983. Science219, 632.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohan Sreelekshmi et al.
Journal of biochemical and molecular toxicology, 35(6), 1-8 (2021-03-03)
Glucokinase (GK), a key regulator of hepatic glucose metabolism in the liver and glucose sensor and mediator in the secretion of insulin in the pancreas, is not studied in detail for its therapeutic application in diabetes. Herein, we study the
Kapil Manglani et al.
Cell biology international, 46(3), 488-497 (2021-12-06)
Role of CDK5 and its inhibition in various neuronal processes and functions are well established. However, role of CDK5 and its inhibition in neuronal insulin-signaling and-resistance is not yet explored. In the present study, we investigated the effect of CDK5
Medha Sharma et al.
Cell communication and signaling : CCS, 20(1), 179-179 (2022-11-16)
The aim of the present study was to determine the role of individual PHLPP isoforms in insulin signaling and insulin resistance in neuronal cells. PHLPP isoforms were either silenced or overexpressed individually, and the effects were observed on individual Akt
Yamini Yadav et al.
The FEBS journal, 289(23), 7561-7581 (2022-07-11)
PP2Cα is one of the newly identified isoforms of metal-dependent protein phosphatases (PPM). The role of this phosphatase in neuronal insulin signalling is completely unknown. In the present study, we show insulin-mediated rapid upregulation of a protein of the insulin
Kerrie A Morrison et al.
Scientific reports, 12(1), 3532-3532 (2022-03-05)
Extraction of integral membrane proteins with poly(styrene-co-maleic acid) provides a promising alternative to detergent extraction. A major advantage of extraction using copolymers rather than detergent is the retention of the lipid bilayer around the proteins. Here we report the first

Questions

  1. I want know about the MW and purity of this product

    1 answer
    1. The molecular weight of Product 407709 Insulin, Zinc, Human, Recombinant, is 5807 Da. Purity is 99.2 % pure by HPLC, < 10.0 EnU/mg, and Potency of 28.5 IU/mg.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service