Skip to Content
Merck
All Photos(1)

Documents

91077C

SAFC

Insulin, Human Recombinant

dry powder, for research or for further manufacturing use

Synonym(s):

Insulin sodium salt human, rh-Insulin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.77

form

dry powder

potency

≥27.5 units per mg

technique(s)

cell culture | mammalian: suitable

impurities

≤1.0 EU/mL endotoxin

solubility

water: 10 mg/mL (pH 2.0—2.5)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

InChI key

PBGKTOXHQIOBKM-FHFVDXKLSA-N

Gene Information

human ... INS(3630)

Looking for similar products? Visit Product Comparison Guide

General description

Insulin, recombinant human (rHu) is identical in function and structure to the native human sequence. A hormone consisting of two polypeptide chains, rHu Insulin′s A-chain (21 amino acids) and B-chain (30 amino acids) are covalently linked by disulfide bonds between cysteine residues. This genetically engineered human insulin is a full sequence molecule producted by recombinant DNA technology in baker′s yeast.

Application

Serum-free medium supplements such as insulin are essential for long-term growth of commonly used mammalian cell lines. When insulin is absent from media, cell may exhibit disturbances in morphology and growth rate.
  • Recombinant Human Insulin has been used in the media for adipogenic induction of mesenchymal stem cells.
  • It has been used as a supplement in the cell culture media of airway epithelial cells.
  • It has been used in the culture media for differentiation of cells, GLUT4 (glucose transporter type 4) translocation assay and to study interaction of insulin with insulin receptor via surface plasmon resonance.

Biochem/physiol Actions

Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Other Notes

InChI: 1S/C257H383N65O77S6/c1-29-131(23)205(313-193(339)104-259)252(393)317-204(130(21)22)248(389)288-159(75-82-200(349)350)217(358)282-156(71-78-189(263)335)221(362)308-183-116-403-404-117-184-243(384)305-178(111-324)240(381)294-162(88-123(7)8)225(366)295-168(95-140-53-61-146(329)62-54-140)228(369)283-154(69-76-187(261)333)218(359)290-161(87-122(5)6)223(364)285-158(74-81-199(347)348)220(361)302-174(101-190(264)336)235(376)298-170(97-142-57-65-148(331)66-58-142)231(372)309-182(242(383)304-176(255(396)397)103-192(266)338)115-402-400-113-180(307-226(367)163(89-124(9)10)291-232(373)172(99-144-105-269-119-275-144)300-219(360)155(70-77-188(262)334)284-234(375)175(102-191(265)337)303-249(390)201(127(15)16)314-211(352)150(260)92-137-44-34-31-35-45-137)213(354)274-109-196(342)280-177(110-323)239(380)301-173(100-145-106-270-120-276-145)233(374)293-165(91-126(13)14)237(378)315-202(128(17)18)247(388)287-157(73-80-198(345)346)215(356)277-133(25)210(351)289-160(86-121(3)4)224(365)296-169(96-141-55-63-147(330)64-56-141)229(370)292-164(90-125(11)12)236(377)316-203(129(19)20)250(391)311-181(114-401-405-118-185(310-244(183)385)245(386)319-207(134(26)326)253(394)306-179(112-325)241(382)318-206(132(24)30-2)251(392)312-184)214(355)273-107-194(340)278-153(72-79-197(343)344)216(357)281-151(51-42-84-271-257(267)268)212(353)272-108-195(341)279-166(93-138-46-36-32-37-47-138)227(368)297-167(94-139-48-38-33-39-49-139)230(371)299-171(98-143-59-67-149(332)68-60-143)238(379)320-208(135(27)327)254(395)322-85-43-52-186(322)246(387)286-152(50-40-41-83-258)222(363)321-209(136(28)328)256(398)399/h31-39,44-49,53-68,105-106,119-136,150-186,201-209,323-332H,29-30,40-43,50-52,69-104,107-118,258-260H2,1-28H3,(H2,261,333)(H2,262,334)(H2,263,335)(H2,264,336)(H2,265,337)(H2,266,338)(H,269,275)(H,270,276)(H,272,353)(H,273,355)(H,274,354)(H,277,356)(H,278,340)(H,279,341)(H,280,342)(H,281,357)(H,282,358)(H,283,369)(H,284,375)(H,285,364)(H,286,387)(H,287,388)(H,288,389)(H,289,351)(H,290,359)(H,291,373)(H,292,370)(H,293,374)(H,294,381)(H,295,366)(H,296,365)(H,297,368)(H,298,376)(H,299,371)(H,300,360)(H,301,380)(H,302,361)(H,303,390)(H,304,383)(H,305,384)(H,306,394)(H,307,367)(H,308,362)(H,309,372)(H,310,385)(H,311,391)(H,312,392)(H,313,339)(H,314,352)(H,315,378)(H,316,377)(H,317,393)(H,318,382)(H,319,386)(H,320,379)(H,321,363)(H,343,344)(H,345,346)(H,347,348)(H,349,350)(H,396,397)(H,398,399)(H4,267,268,271)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tracey Pirali et al.
ChemMedChem, 12(18), 1542-1554 (2017-09-01)
Activation of the phosphoinositide 3-kinase (PI3K) pathway is a key signaling event in cancer, inflammation, and other proliferative diseases. PI3K inhibitors are already approved for some specific clinical indications, but their systemic on-target toxicity limits their larger use. In particular
Glucocorticoids Distinctively Modulate the CFTR Channel with Possible Implications in Lung Development and Transition into Extrauterine Life.
Laube M, et al.
PLoS ONE, 10, e0124833-e0124833 (2015)
Senthil Raja Jayapal et al.
Cell cycle (Georgetown, Tex.), 15(22), 3070-3081 (2016-09-23)
Cyclin A2 is an essential gene for development and in haematopoietic stem cells and therefore its functions in definitive erythropoiesis have not been investigated. We have ablated cyclin A2 in committed erythroid progenitors in vivo using erythropoietin receptor promoter-driven Cre
Guokun Yang et al.
General and comparative endocrinology, 281, 137-144 (2019-06-10)
The insulin-like growth factor (IGF) system plays a pivotal role in the regulation of growth, and IGF binding proteins (IGFBPs) are important regulatory factors in the IGF system. Generally, IGFBPs inhibit IGF actions by preventing its binding to receptors. Under
Xiu Min Ang et al.
Tissue engineering. Part A, 20(5-6), 966-981 (2013-10-24)
The microenvironment plays a vital role in both the maintenance of stem cells in their undifferentiated state (niche) and their differentiation after homing into new locations outside this niche. Contrary to conventional in-vitro culture practices, the in-vivo stem cell microenvironment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service