Skip to Content
Merck
All Photos(1)

Key Documents

I3536

Sigma-Aldrich

Insulin human

recombinant, expressed in yeast, γ-irradiated, suitable for cell culture

Synonym(s):

Insulin human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C257H383N65O77S6
CAS Number:
Molecular Weight:
5807.57
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77
biological source:
Saccharomyces cerevisiae
form:
lyophilized powder
Pricing and availability is not currently available.

biological source

Saccharomyces cerevisiae

Quality Level

recombinant

expressed in yeast

sterility

γ-irradiated

form

lyophilized powder

potency

≥25 USP units per mg

technique(s)

cell culture | mammalian: suitable

solubility

0.01 M HCl: 20 mg/mL, clear, colorless to faintly yellow

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

InChI key

PBGKTOXHQIOBKM-FHFVDXKLSA-N

Gene Information

human ... INS(3630)

Looking for similar products? Visit Product Comparison Guide

General description

The INS gene encodes for preproinsulin, which is enzymatically converted into insulin.[1] Insulin is produced in the insulin-producing pancreatic β cells.[2] Preproinsulin is converted to proinsulin in ER and proinsulin is then proteolytically processed to form insulin in newly-forming insulin secretory granules.[3] Insulin production is tightly regulated by specific DNA elements present within ∼400 bp in the proximal region of the INS promoter.[4]

Application

Insulin human has been used for the following applications:
  • For adipogenic differentiation assays (in the preparation of medium supplement)[5]
  • Incubation of cells for the evaluation of the effects of insulin[6]
  • Mass spectrometry (used for the external caliberation)[7]

Biochem/physiol Actions

Insulin is responsible for two types of actions- excitatory and inhibitory. In its excitatory role, it increases the uptake of glucose and lipid synthesis, and in its inhibitory role it inhibits glycogenolysis, gluconeogenesis, lipolysis, proteolysis and ketogenesis.[8] Aberrant insulin secretion leads to various disorders such as diabetes, hyperglycemia or hypoglycaemia.[2] Type I diabetes is a result of autoimmune destruction of β cells of pancreas, which leads to depletion of insulin.[9] Mutant INS-gene Induced Diabetes of Youth (MIDY) syndrome is an autosomal dominant disorder caused by missense mutations, which lead to aberrant proinsulin folding.[3] Impaired glucose tolerance (IGT) or non-insulin-dependent diabetes mellitus (NIDDM) is caused by resistance to insulin-stimulated glucose uptake.[10]
Regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.
Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Other Notes

InChI: 1S/C257H383N65O77S6/c1-29-131(23)205(313-193(339)104-259)252(393)317-204(130(21)22)248(389)288-159(75-82-200(349)350)217(358)282-156(71-78-189(263)335)221(362)308-183-116-403-404-117-184-243(384)305-178(111-324)240(381)294-162(88-123(7)8)225(366)295-168(95-140-53-61-146(329)62-54-140)228(369)283-154(69-76-187(261)333)218(359)290-161(87-122(5)6)223(364)285-158(74-81-199(347)348)220(361)302-174(101-190(264)336)235(376)298-170(97-142-57-65-148(331)66-58-142)231(372)309-182(242(383)304-176(255(396)397)103-192(266)338)115-402-401-114-181(214(355)273-107-194(340)278-153(72-79-197(343)344)216(357)281-151(51-42-84-271-257(267)268)212(353)272-108-195(341)279-166(93-138-46-36-32-37-47-138)227(368)297-167(94-139-48-38-33-39-49-139)230(371)299-171(98-143-59-67-149(332)68-60-143)238(379)320-208(135(27)327)254(395)322-85-43-52-186(322)246(387)286-152(50-40-41-83-258)222(363)321-209(136(28)328)256(398)399)311-250(391)203(129(19)20)316-236(377)164(90-125(11)12)292-229(370)169(96-141-55-63-147(330)64-56-141)296-224(365)160(86-121(3)4)289-210(351)133(25)277-215(356)157(73-80-198(345)346)287-247(388)202(128(17)18)315-237(378)165(91-126(13)14)293-233(374)173(100-145-106-270-120-276-145)301-239(380)177(110-323)280-196(342)109-274-213(354)180(113-400-405-118-185(310-244(183)385)245(386)319-207(134(26)326)253(394)306-179(112-325)241(382)318-206(132(24)30-2)251(392)312-184)307-226(367)163(89-124(9)10)291-232(373)172(99-144-105-269-119-275-144)300-219(360)155(70-77-188(262)334)284-234(375)175(102-191(265)337)303-249(390)201(127(15)16)314-211(352)150(260)92-137-44-34-31-35-45-137/h31-39,44-49,53-68,105-106,119-136,150-186,201-209,323-332H,29-30,40-43,50-52,69-104,107-118,258-260H2,1-28H3,(H2,261,333)(H2,262,334)(H2,263,335)(H2,264,336)(H2,265,337)(H2,266,338)(H,269,275)(H,270,276)(H,272,353)(H,273,355)(H,274,354)(H,277,356)(H,278,340)(H,279,341)(H,280,342)(H,281,357)(H,282,358)(H,283,369)(H,284,375)(H,285,364)(H,286,387)(H,287,388)(H,288,389)(H,289,351)(H,290,359)(H,291,373)(H,292,370)(H,293,374)(H,294,381)(H,295,366)(H,296,365)(H,297,368)(H,298,376)(H,299,371)(H,300,360)(H,301,380)(H,302,361)(H,303,390)(H,304,383)(H,305,384)(H,306,394)(H,307,367)(H,308,362)(H,309,372)(H,310,385)(H,311,391)(H,312,392)(H,313,339)(H,314,352)(H,315,378)(H,316,377)(H,317,393)(H,318,382)(H,319,386)(H,320,379)(H,321,363)(H,343,344)(H,345,346)(H,347,348)(H,349,350)(H,396,397)(H,398,399)(H4,267,268,271)/t131-,132-,133-,134+,135+,136+,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,186-,201-,202-,203-,204-,205-,206-,207-,208-,209-/m0/s1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nakamura et al.
British journal of pharmacology, 123(4), 675-682 (1998-03-28)
1. The actions of troglitazone, pioglitazone, metformin and bezafibrate, agents that improve insulin-resistance, on voltage-dependent Ca2+ channels in arterial smooth muscle cells were examined by use of the conventional and nystatin-perforated whole-cell clamp methods. Single cells were freshly isolated from
E Roine et al.
FEBS letters, 417(2), 168-172 (1997-12-12)
Pseudomonas syringae pv. tomato DC3000 produces Hrp pili under inducing in vitro conditions. A preparation of partially purified extracellular filaments contains HrpA, flagellin and some minor contaminants. HrpA was separated from the major contaminant, the flagellin, by gel filtration to
Adeline C Y Chua et al.
Biomaterials, 216, 119221-119221 (2019-06-14)
Hypnozoites are the liver stage non-dividing form of the malaria parasite that are responsible for relapse and acts as a natural reservoir for human malaria Plasmodium vivax and P. ovale as well as a phylogenetically related simian malaria P. cynomolgi.
Swarup K Chakrabarti et al.
The Journal of biological chemistry, 278(26), 23617-23623 (2003-04-25)
Histone modifying enzymes contribute to the activation or inactivation of transcription by ultimately catalyzing the unfolding or further compaction, respectively, of chromatin structure. Actively transcribed genes are typically hyperacetylated at Lys residues of histones H3 and H4 and hypermethylated at
G M Reaven
Diabetes, 37(12), 1595-1607 (1988-12-01)
Resistance to insulin-stimulated glucose uptake is present in the majority of patients with impaired glucose tolerance (IGT) or non-insulin-dependent diabetes mellitus (NIDDM) and in approximately 25% of nonobese individuals with normal oral glucose tolerance. In these conditions, deterioration of glucose

Questions

  1. After reconstitution in 0.01 M HCl, can the aliquot be frozen and for how long?

    1 answer
    1. Insulin in solution is reasonably stable. Sterile solutions can be stored at 2–8 °C for up to 12 months. Should freezing be preferred, solutions may be stored frozen at -20°C in single-use aliquots. Freeze-thaw cycles should be avoided.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service