1.03053
N,N-Dimethylformamide
for analysis EMSURE® ACS,ISO,Reag. Ph Eur
Synonym(s):
DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. ISO
reag. Ph. Eur.
vapor density
2.5 (vs air)
vapor pressure
2.7 mmHg ( 20 °C)
product line
EMSURE®
Assay
≥99.8% (GC)
form
liquid
autoignition temp.
833 °F
potency
3010 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)
expl. lim.
15.2 %
impurities
≤0.00005% Al (Aluminium)
≤0.0005 meq/g Titrable acid
≤0.003 meq/g Titrable base
≤0.1% Water
evapn. residue
≤0.001%
color
clearAPHA: ≤10
refractive index
n20/D 1.430 (lit.)
pH
7 (20 °C, 200 g/L in H2O)
bp
153 °C (lit.)
mp
−61 °C (lit.)
transition temp
flash point 57.5 °C
solubility
soluble 1000 g/L
density
0.944 g/mL (lit.)
cation traces
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
SMILES string
[H]C(=O)N(C)C
storage temp.
2-30°C
InChI
1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3
InChI key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Titrable acid: ≤ 0.0005 meq/g
Titrable base: ≤ 0.003 meq/g
Density (d 20 °C/20 °C): 0.949 - 0.952
Refractive index (n 20/D): 1.429 - 1.431
Boiling point: 152 - 154 °C
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.10 %
Legal Information
greener alternative product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
135.5 °F - closed cup
Flash Point(C)
57.5 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service