Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N,N
-Dimethylformamide
Synonym(s):
DMF, NSC 5356,
N,N
-Dimethylformamide
Linear Formula:
HCON(CH
3
)
2
CAS Number:
68-12-2
Molecular Weight:
73.09
EC Number:
200-679-5
Products
Product Category
solvents (17)
Brand
Sigma-Aldrich (8)
Supelco (11)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (2)
Anhydrous (2)
Biotechnology (1)
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
319937
N,N
-Dimethylformamide,
ACS reagent, ≥99.8%
Expand
View Pricing
270547
N,N
-Dimethylformamide,
suitable for HPLC, ≥99.9%
Expand
View Pricing
5.89565
N,N
-Dimethylformamide,
≥99.8%, anhydrous
Expand
View Pricing
1.03053
N,N
-Dimethylformamide,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
L091000
N,N
-Dimethylformamide,
Expand
View Pricing
D158550
N,N
-Dimethylformamide,
ReagentPlus
®
, ≥99%
Expand
View Pricing
D4551
N,N
-Dimethylformamide,
Molecular Biology, ≥99%
Expand
View Pricing
1.00202
N,N
-Dimethylformamide,
for headspace gas chromatography SupraSolv
®
Expand
View Pricing
494488
N,N
-Dimethylformamide,
biotech. grade, ≥99.9%
Expand
View Pricing
1.00397
N,N
-Dimethylformamide,
for peptide synthesis
Expand
View Pricing
8.22275
N,N
-Dimethylformamide,
EMPLURA
®
Expand
View Pricing
1.10983
N,N
-Dimethylformamide,
for gas chromatography ECD and FID SupraSolv
®
Expand
View Pricing
1.03034
N,N
-Dimethylformamide,
EMPARTA
®
Expand
View Pricing
5.43897
N,N
-Dimethylformamide,
for liquid chromatography LiChrosolv
®
Expand
View Pricing
PHR1553
N,N
-Dimethylformamide,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
DX1730
N,N
-Dimethylformamide,
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
1.02937
N,N
-Dimethylformamide,
for spectroscopy Uvasol
®
Expand
View Pricing
900638
N,N
-Dimethylformamide,
anhydrous, ZerO
2
®
, 99.8%
Expand
View Pricing
DX1729
N,N
-Dimethylformamide,
Expand
View Pricing
72438
N,N
-Dimethylformamide,
analytical standard
Expand
View Pricing
Page 1 of 2
Page 1 of 2