Skip to Content
Merck
All Photos(12)

Key Documents

1.00983

Supelco

Ethanol

greener alternative

absolute for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Ethyl alcohol, Methylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-578-6
NACRES:
NB.21
grade:
ACS reagent
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
78 °C (lit.)
vapor pressure:
59 hPa ( 20 °C)

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

1.59 (vs air)

vapor pressure

59 hPa ( 20 °C)

description

for analytical purposes
for extraction

product line

EMSURE®

Assay

≥99.9% (GC)

form

liquid

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

24.5 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

dilution

(for analytical testing)

refractive index

n20/D 1.36 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−130 (lit.)

transition temp

flash point 12 °C

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

greener alternative category

storage temp.

2-30°C

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biorenewable and alternative to petrochemical solvents. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here  for more information.

Application

Ethanol has been used as a solvent in the synthesis of Pd/SiO2 xerogel catalysts for hydrodechlorination of dichloroethane.

Features and Benefits

Our bioethanol is an obvious choice for a sustainable future because of its high quality, affordability, and ready availability.      
  • It is produced from grain or sugar cane, a renewable source
  • Less toxic than synthetic ethanol (no toxic by-products)
  • Reliable availability      
  • Production method is safer for the environment

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Jae Ho Choi et al.
Cells, 8(12) (2019-12-05)
Platycodi radix is a widely sold health food worldwide, which contains numerous phytochemicals that are beneficial to health. Previously, we reported that saponin from the roots of Platycodi radix-derived saponin inhibited toxicant-induced liver diseases. Nevertheless, the inhibitory effect of platyconic
Improvement of metal dispersion in Pd/SiO2 cogelled xerogel catalysts for 1, 2-dichloroethane hydrodechlorination
Lambert S, et al.
Applied Catalysis. B, Environmental, 50(2), 127-140 (2004)
Jueun Kim et al.
Journal of microbiology (Seoul, Korea), 58(1), 11-16 (2019-06-13)
A chromatin immunoprecipitation (ChIP) assay is a method to identify how much a protein of interest binds to the DNA region. This method is indispensable to study the mechanisms of how the transcription factors or chromatin modifications regulate the gene
Anita Yadav et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 19(2), 251-260 (2020-01-28)
The effects of ionic liquid addition on the spectroscopic properties of a pyrene-tryptophan-containing fluorescent intramolecular complex in polar-aprotic and polar-protic solvents, specifically, acetonitrile and ethanol, are assessed. Two ionic liquid sets, consisting of seven different ionic liquids, were explored; set
Yun Hee Kim et al.
Journal of medicinal food, 23(5), 491-498 (2020-03-19)
Changing consumption patterns and increasing health awareness, especially in Europe, are resulting in an increased demand for sesame seeds. In 2016, Asia imported the highest quantity of sesame seeds, followed by Europe and North America. We examined, for the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service