Skip to Content
Merck
All Photos(6)

Documents

920193

Sigma-Aldrich

Cyrene 2-Methyltetrahydrofuran Blend

greener alternative

BioRenewable, DMF and NMP Substitute

Synonym(s):

2-MeTHF Solution, 2-Methyltetrahydrofuran solution, Cyrene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
Molecular Weight:
128.13
UNSPSC Code:
12352212
NACRES:
NA.07

form

liquid

contains

30-80 ppm BHT as stabilizer

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

renewable carbon content

100%

density

1.153 g/mL

greener alternative category

General description

Cyrene 2-Methyltetrahydrofuran Blend with BHT as an inhibitor is specially formulated for use in automated processes and applications where lower viscosity is required, making it easier to handle while maintaining high yields.

This Cyrene blend is an alternative for NMP and DMF in many cross-coupling reactions. Neither Cyrene nor 2-Methyltetrahydrofuran have any genotoxic or mutagenic concerns associated with their use, making them much safer alternatives to traditional aprotic solvents.

This new blend is part of our Biorenewable product line, meaning that it is made from renewable resources and does not compete with food resources for its production, making it more environmentally sustainable for the future. The material has also been independently certified as 100% renewable carbon.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is BioRenewable and thus aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here for more information.

Application

This cosolvent solution has been shown to have similar reaction yields to NMP and DMF for many cross-coupling reactions, such as:

Features and Benefits

  • Independently certified as 100% renewable carbon.
  • Closer physical properties of NMP and DMF while maintaining high reaction yields.
  • Sustainably produced and environmentally less hazardous than more traditional petroleum aprotic solvents
  • Both Cyrene and 2-MeTHF are also much safer for handling without the genotoxic concerns of other solvents.
  • It has a lower viscosity than neat Cyrene making it more optimized for automated processes.

Legal Information

CYRENE is a trademark of Circa Group Pvt Ltd

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved 2-pyridyl reductive homocoupling reaction using biorenewable solvent Cyrene?(dihydrolevoglucosenone)
Webb DA, et al.
RSC Sustainability, 1, 1522-1529 (2023)
Cyrene? blends: a greener solvent system for organic syntheses
Sullivan C, et al.
Green Chemistry, 24, 7184-7193 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service