Skip to Content
Merck
All Photos(1)

Key Documents

72438

Supelco

N,N-Dimethylformamide

analytical standard

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

833 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

15.2 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.2% water
water

refractive index

n20/D 1.430 (lit.)
n20/D 1.430

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide is an non-volatile,[1] organic solvent, which finds applications in chemical industries as a solvent, additive and intermediate.[2]

Application

N,N-Dimethylformamide has been used as a solvent in determining the lipase activity of the crude protein extracts separated from different plant tissues using sodium dodecyl sulfate (SDS) page assay technique.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Solvent for many hydrophobic organic compounds.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Gel Detection of Lipase Activity in Crude Plant Extracts (Olea europaea).
Zienkiewicz A, et al.
Bio-protocol (2015)
Upper critical solution temperature behavior of poly (N-isopropylacrylamide) in an ionic liquid and preparation of thermo-sensitive nonvolatile gels.
Takeshi U and Masayoshi W.
Chemistry Letters (Jpn), 35(8), 964-965 (2006)
N,N-dimethylformamide, Issue 31 (2001)
Kun Liu et al.
Nanoscale, 4(20), 6574-6580 (2012-09-15)
Studies on the self-assembly of metal nanoparticles (NPs) in the presence of ions are motivated by the biosensing applications of NP clusters and the capability to control the morphology of clusters of oppositely charged NPs. The effect of ions has
L Piazza et al.
Nature communications, 6, 6407-6407 (2015-03-03)
Surface plasmon polaritons can confine electromagnetic fields in subwavelength spaces and are of interest for photonics, optical data storage devices and biosensing applications. In analogy to photons, they exhibit wave-particle duality, whose different aspects have recently been observed in separate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service