1.00030
Acetonitrile
≥99.9% (GC), gradient grade, suitable for HPLC, Reag. Ph Eur, LiChrosolv®
Synonym(s):
ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide
About This Item
Recommended Products
product name
Acetonitrile, gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur
grade
gradient grade
Quality Level
Agency
reag. Ph. Eur.
vapor density
1.41 (vs air)
vapor pressure
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)
product line
LiChrosolv®
Assay
≥99.9% (GC)
form
liquid
autoignition temp.
524 °C
973 °F
expl. lim.
16 %
technique(s)
HPLC: suitable
impurities
≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water
evapn. residue
≤2.0 mg/L
color
APHA: ≤10
transmittance
193 nm, ≥60%
195 nm, ≥80%
230 nm, ≥98%
refractive index
n20/D 1.344 (lit.)
bp
81-82 °C (lit.)
mp
−45 °C (lit.)
transition temp
flash point 2 °C
density
0.786 g/mL at 25 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
CC#N
InChI
1S/C2H3N/c1-2-3/h1H3
InChI key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- A solvent for the mobile phase during the solid phase extraction (SPE) for the preconcentration of Bisphenol A (BPA) from tea samples.
- An extraction solvent for volatile organic compounds from concentrated indoor air samples.
- A rinsing solvent for glassware.
Preparation Note
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service